Stopper Assembly
New Product
 :   SAVILLEX
Stopper Assembly
 :  SAVILLEX
New Product
Stopper Assembly
  • For use in glass flasks
    Ferrule nut accepts ³/₈" OD tube
Product Family Options
  • Désignation
    Taille
    For Use With
  • Stopper assembly, #6 taper
    30 mm
    Each
    Each - $125.56
    $125.56
    Each
       $125.56
     : 730-0066
    New Product
    New Product
  • Stopper assembly, #8 taper
    37.3 mm
    For glass flasks
    Each
    Each - $125.56
    $125.56
    Each
       $125.56
     : 730-0086