Closures, PFA
New Product
Supplier: SAVILLEX
Jump to:
À propos de cet article
Service temperature range of PFA is ‑200°C to 260°C (‑328°F to 500°F)
Knurled finish on the closure
Leakproof seal
Push‑in ports allow for 'closed seal' drying on hot plate
2 Options Available Below
Product Details & Documents
Service temperature range of PFA is ‑200°C to 260°C (‑328°F to 500°F)
Knurled finish on the closure
Leakproof seal
Push‑in ports allow for 'closed seal' drying on hot plate
Product Family Options
Informations sur le produit
- DésignationTailleMatièreTaille du bouchonAvailabilityPrice
- Supplier Number: 600-033-23Spécifications:New ProductNew Product
- Supplier Number: 600-033-71Spécifications:New ProductNew Product