Recessed Closures
New Product
 :   SAVILLEX
Recessed Closures
 :  SAVILLEX
New Product
Recessed Closures
Closures are molded PFA and are available for all Savillex products designed to accept a closure. Transfer closures come complete with ferrule nuts on the tube ports. A wrench set is recommended to ensure a tight fit and best seal for all products where a wrench set is available.
  • Service temperature range of PFA is –200 to 260 °C (–328 to 500 °F)
    External grips on the closure fit 110 mm wrench
    Recessed area for Nomex write on tabs
    Leakproof seal
Product Family Options
  • Désignation
    Taille du bouchon
    Matière
    Cap Type
  • Recessed closure
    53 mm
    PFA
    Recessed
    Each
    Each - 66,12 $
    66,12 $
    Each
       66,12 $
     : 600-053-71
    New Product
    New Product
  • Recessed closure
    70 mm
    PFA
    Recessed
    Each
    Each - 62,60 $
    62,60 $
    Each
       62,60 $
     : 600-070-71
    New Product
    New Product
  • Recessed closure
    89 mm
    PFA
    Recessed
    Each
    Each - 91,88 $
    91,88 $
    Each
       91,88 $
     : 600-089-71
    New Product
    New Product
  • Recessed closure
    110 mm
    PFA
    Recessed
    Each
    Each - 171,25 $
    171,25 $
    Each
       171,25 $
     : 600-110-71