Recessed Closures
Closures are molded PFA and are available for all Savillex products designed to accept a closure. Transfer closures come complete with ferrule nuts on the tube ports. A wrench set is recommended to ensure a tight fit and best seal for all products where a wrench set is available.
Service temperature range of PFA is –200 to 260 °C (–328 to 500 °F)
External grips on the closure fit 110 mm wrench
Recessed area for Nomex write on tabs
Leakproof seal
Closures are molded PFA and are available for all Savillex products designed to accept a closure. Transfer closures come complete with ferrule nuts on the tube ports. A wrench set is recommended to ensure a tight fit and best seal for all products where a wrench set is available.
Service temperature range of PFA is –200 to 260 °C (–328 to 500 °F)
External grips on the closure fit 110 mm wrench
Recessed area for Nomex write on tabs
Leakproof seal
Product Family Options
- DésignationTaille du bouchonMatièreCap Type
- : 600-053-71New ProductNew Product
- : 600-070-71New ProductNew Product
- : 600-089-71New ProductNew Product
- : 600-110-71New ProductNew Product