VWR® Basic Protection Scrub Apparels
VWR® Basic Protection Scrub Apparels
VWR® Basic Protection Scrub Apparels
Supplier:  VWR International
Certificates
6 Options Available Below
Product Details & Documents
Comfortable and practical scrub apparel designed for cleanroom and laboratory environments. Made from lightweight, breathable material, these scrub pants and shirts provide essential coverage and durability for everyday use. They feature a simple design for ease of movement and provide cost-effective protection in controlled settings.
  • Lightweight, breathable material is made from non-woven fabric for comfort during extended wear
  • Two-piece design includes scrub top and pants (sold separately) for full coverage and mobility
  • Elastic waistband and adjustable ties ensure a secure and comfortable fit
  • Disposable, single-use design reduces contamination risk and eliminates laundering
  • Latex-free garments are safe for individuals with latex sensitivities
  • Suitable for general cleanroom, lab, and healthcare environments
  • Helps maintain cleanliness and safety standards in controlled environments
Basic Protection Scrub apparel offers practical, comfortable coverage for cleanroom, laboratory, and healthcare environments. Made from lightweight, breathable material, these disposable scrubs include a two-piece design with a top and pants for full mobility and ease of wear. Featuring elastic waistbands and adjustable ties, they provide a secure fit while supporting contamination control. Ideal for non-sterile applications, Basic Protection Scrubs deliver cost-effective protection without sacrificing comfort or compliance.
Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Packaging: Garments are packaged 30 pieces per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89235-856
  • 89235-858
  • 89235-860
  • 89235-862
  • 89235-864
  • 89235-866
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Weight:
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
Specifications
Product Family Options
Scrub shirts
Product Information
  • Color
    Taille de confection
    Longueur
    Chest Size
    Waist Size
    Inseam
    Sleeve
    Availability
    Price
  • Dark blue
    L
    28"
    27"
    27½"
    20"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $122.26
    $122.26
    New Customer Promotional Price
    Case of 30
       $122.26
    Specifications:
    More
    Specifications
    Cat. No.
    89235-856
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²
  • Dark blue
    XL
    29½"
    28"
    28½"
    21"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $125.05
    $125.05
    New Customer Promotional Price
    Case of 30
       $125.05
    Specifications:
    More
    Specifications
    Cat. No.
    89235-858
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²
  • Dark blue
    XXL
    30"
    30"
    30½"
    23"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $128.65
    $128.65
    New Customer Promotional Price
    Case of 30
       $128.65
    Specifications:
    More
    Specifications
    Cat. No.
    89235-860
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²
Scrub pants
Product Information
  • Color
    Taille de confection
    Longueur
    Chest Size
    Waist Size
    Inseam
    Sleeve
    Availability
    Price
  • Dark blue
    L
    43"
    21"
    29"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $150.16
    $150.16
    New Customer Promotional Price
    Case of 30
       $150.16
    Specifications:
    More
    Specifications
    Cat. No.
    89235-862
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²
  • Dark blue
    XL
    45"
    22½
    30"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $160.70
    $160.70
    New Customer Promotional Price
    Case of 30
       $160.70
    Specifications:
    More
    Specifications
    Cat. No.
    89235-864
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²
  • Dark blue
    XXL
    46"
    23"
    30"
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $167.72
    $167.72
    New Customer Promotional Price
    Case of 30
       $167.72
    Specifications:
    More
    Specifications
    Cat. No.
    89235-866
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom
    Weight
    50 g/m²