Sealed Jars
New Product
Supplier: SAVILLEX
Jump to:
About this item
Service temperature range of PFA is –328 to +500 °F (–200 to +260 °C)
Molded drain tube in bottom of jar
All sealed jars have two connecting ports
Heavy walled construction good for pressure applications
Pressure rating is for ambient conditions
6 Options Available Below
Product Details & Documents
Service temperature range of PFA is –328 to +500 °F (–200 to +260 °C)
Molded drain tube in bottom of jar
All sealed jars have two connecting ports
Heavy walled construction good for pressure applications
Pressure rating is for ambient conditions
Product Family Options
Product Information
- DésignationMatièreCapacitéHauteurAvailabilityPrice
- Supplier Number: 102-0010-01Specifications:New ProductNew Product
- Supplier Number: 102-0125-01Specifications:New ProductNew Product
- Supplier Number: 102-0475-01Specifications:New ProductNew Product
- Supplier Number: 102-0475-03Specifications:New ProductNew Product
- Supplier Number: 102-1100-01Specifications:New ProductNew Product
- Sealed jar with molded drain with 2×¼" portsPFA1100 ml163.8 mmLog in for availabilityEach$1,107.00Supplier Number: 102-1100-03Specifications:New ProductNew Product