Stopper Assembly
Stopper Assembly, 33 mm, #7 Taper
78473-872
New Product
: SAVILLEX
: 730-0076
The stopper assembly is molded from PFA, with a service temperature range of −200 to +260 °C. It is designed to fit standard tapers of laboratory glassware and serves as a connector between Savillex filter assemblies and glass flasks
- Désignation:Stopper Assembly, 33 mm
- For Use With:#7 Taper