Column Component Closures
New Product
 :   SAVILLEX
Column Component Closures
 :  SAVILLEX
New Product
Column Component Closures
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Product Family Options
  • Désignation
  • Column component closure, with one ¹/₄" FNPT port, 58 mm
    Each
    Each - $102.53
    $102.53
    Each
       $102.53
     : 600-058-17
    New Product
    New Product
  • Column component closure, with one ³/₈" FNPT port, 58 mm
    Each
    Each - $120.46
    $120.46
    Each
       $120.46
     : 600-058-19
    New Product
    New Product
  • Column component closure, with two ¹/₈" outer Ø tube port, 58 mm
    Each
    Each - $136.91
    $136.91
    Each
       $136.91
     : 600-058-24
    New Product
    New Product
  • Column component closure, with two ¹/₄" outer Ø tube port, 58 mm
    Each
    Each - $155.90
    $155.90
    Each
       $155.90
     : 600-058-33
    New Product
    New Product
  • Column component closure, with two ¹/₈" push in transfer side ports, 24 mm
    Each
    Each - $56.90
    $56.90
    Each
       $56.90
     : 600-024-23
    New Product
    New Product
  • Column component closure, 58 mm
    Each
    Each - $56.97
    $56.97
    Each
       $56.97
     : 600-058-06
    New Product
    New Product
  • Column component closure, with one ¹/₄" outer Ø tube port, 58 mm
    Each
    Each - $87.42
    $87.42
    Each
       $87.42
     : 600-058-16
    New Product
    New Product
  • Column component closure, with two ¹/₂" outer Ø tube port, 58 mm
    Each
    Each - $226.32
    $226.32
    Each
       $226.32
     : 600-058-42
    New Product
    New Product
  • Transfer closure, pressure relief valve, Ultem® clamp, 1¹/₂" FNPT threads, 58 mm
    Each
    Each - $418.79
    $418.79
    Each
       $418.79
     : 600-058-87