Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cleaning Container Lid

Cleaning Container Lid

Cleaning container lid, length: 60.3 mm, Ø 166.4 mm
Catalog # 78474-281
New Product
Supplier:   SAVILLEX
Supplier #:  550-740-400817
Cleaning Container Lid
Catalog # 78474-281
Supplier:  SAVILLEX
New Product
Cleaning Container Lid

Cleaning container lid, length: 60.3 mm, Ø 166.4 mm

Price
$279.44
Log in for availability
Each
Product Details & Documents

Fits cleaning container. ½" and ¼" tube port. Fittings not included.

½" tube port only accepts 78473-822 or 78474-135. Requires sealing ring 78474-021.

Specifications
  • Désignation:
    Cleaning container lid, length: 60.3 mm, Ø 166.4 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Pour spout assembly
    Pour spout assembly
    78474-475
    Log in for availability
    Each - $73.33

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.