Syringes, PFA
À propos de cet article
Ideal for ultrapure liquid transfer in environmental, biomedical and electronics applications.
Molded PFA syringes are available with a 10 or 20 ml capacity
PFA is chemically inert and resistant to temperature extremes
Integral plunger is made of high purity PTFE, with no O-rings or external seals required.
Service temperature range of PFA is −200 to +260 °C (−328 to 500 °F)
5 Options Available Below
Product Details & Documents
Ideal for ultrapure liquid transfer in environmental, biomedical and electronics applications.
Molded PFA syringes are available with a 10 or 20 ml capacity
PFA is chemically inert and resistant to temperature extremes
Integral plunger is made of high purity PTFE, with no O-rings or external seals required.
Service temperature range of PFA is −200 to +260 °C (−328 to 500 °F)
Product Family Options
Syringes
Informations sur le produit
- DésignationCapacitéLongueurØ ext.AvailabilityPrice
- Syringe with female leur‑lock attachment, used with 13 mm filters and dispensing tube10 ml103.1 mm (Length), 118.5 mm (Overall length)19.7 mmLog in pour la disponibilitéEach$276.97Supplier Number: 700-510Spécifications:New ProductNew Product
- Syringe with tube connector20 ml115.6 mm (Length), 146.3 mm (Overall length)22.9 mmLog in pour la disponibilitéEach$332.37Supplier Number: 700-520Spécifications:New ProductNew Product
- Syringe with permanent retrieve-dispense tube; 1.5×20 cm (int.Ø×L) (Dispensing tube)20 ml115.6 mm (Length), 349.2 mm (Overall length)22.9 mmLog in pour la disponibilitéEach$459.15Supplier Number: 700-521Spécifications:New ProductNew Product
Syringe tubes
Informations sur le produit
- DésignationCapacitéLongueurØ ext.AvailabilityPrice
- Supplier Number: 700-511Spécifications:New ProductNew Product
- Syringe Push-In Tube for 10 ml syringe, 8Ø×100L mm, used on 13 mm filter for dispensing filtered materialLog in pour la disponibilitéEach$15.97Supplier Number: 700-513Spécifications:New ProductNew Product