Syringes, PFA | Avantor
Syringes, PFA
New Product
Supplier:   SAVILLEX
Syringes, PFA
Supplier:  SAVILLEX
New Product
Certificates
5 Options Available Below
Product Details & Documents
Ideal for ultrapure liquid transfer in environmental, biomedical and electronics applications.
  • Molded PFA syringes are available with a 10 or 20 ml capacity
    PFA is chemically inert and resistant to temperature extremes
Integral plunger is made of high purity PTFE, with no O-rings or external seals required.
Service temperature range of PFA is −200 to +260 °C (−328 to 500 °F)
Product Family Options
Syringes
Informations sur le produit
  • Désignation
    Capacité
    Longueur
    Ø ext.
    Availability
    Price
  • Syringe with female leur‑lock attachment, used with 13 mm filters and dispensing tube
    10 ml
    103.1 mm (Length), 118.5 mm (Overall length)
    19.7 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $276.97
    $276.97
    Each
       $276.97
    Supplier Number: 700-510
    Spécifications:
    New Product
    New Product
  • Syringe with tube connector
    20 ml
    115.6 mm (Length), 146.3 mm (Overall length)
    22.9 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $332.37
    $332.37
    Each
       $332.37
    Supplier Number: 700-520
    Spécifications:
    New Product
    New Product
  • Syringe with permanent retrieve-dispense tube; 1.5×20 cm (int.Ø×L) (Dispensing tube)
    20 ml
    115.6 mm (Length), 349.2 mm (Overall length)
    22.9 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $459.15
    $459.15
    Each
       $459.15
    Supplier Number: 700-521
    Spécifications:
    New Product
    New Product
Syringe tubes
Informations sur le produit
  • Désignation
    Capacité
    Longueur
    Ø ext.
    Availability
    Price
  • Syringe dispense tube for 10 ml syringe, male leur lock
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $113.33
    $113.33
    Each
       $113.33
    Supplier Number: 700-511
    Spécifications:
    New Product
    New Product
  • Syringe Push-In Tube for 10 ml syringe, 8Ø×100L mm, used on 13 mm filter for dispensing filtered material
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.97
    $15.97
    Each
       $15.97
    Supplier Number: 700-513
    Spécifications:
    New Product