Standard Vials
New Product
Supplier:   SAVILLEX
Standard Vials
Supplier:  SAVILLEX
New Product
Certificates
13 Options Available Below
Product Details & Documents
For 40 years, Savillex PFA vials have been the best choice for demanding trace metal applications, and are used in laboratories worldwide for sample preparation and storage.
  • Used typically for storage and also digestion in geochemistry
    Flat base for maximum heating efficiency
    Select the interior shape to suit the application
    Square exterior vials with hex closures have higher pressure rating
    Fin exterior vial used for microdistillation
    Maximum service temperature of PFA is 260 ºC/500 ºF
The bottom type and high service temperature range makes them ideal for use directly on a hot plate. Molded from high purity virgin PFA, they are chemically inert, with ultra low trace metal content.
Product Family Options
Product Information
  • Désignation
    Capacité
    Diameter ×H
    Matière
    Avec bouchon
    Taille du bouchon
    Availability
    Price
  • Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    22 ml
    30.5×55.6 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $43.26
    $43.26
    Each
       $43.26
    Supplier Number: 200-022-12
    Specifications:
    New Product
    New Product
  • Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    22 ml
    30.5×55.6 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $46.86
    $46.86
    Each
       $46.86
    Supplier Number: 200-022-30
    Specifications:
    New Product
    New Product
  • Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    30 ml
    30.5×69.9 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $58.55
    $58.55
    Each
       $58.55
    Supplier Number: 200-030-12
    Specifications:
    New Product
    New Product
  • Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    30 ml
    30.5×79.0 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $52.41
    $52.41
    Each
       $52.41
    Supplier Number: 200-030-30
    Specifications:
    New Product
    New Product
  • Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    60 ml
    30.7×133.9 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $101.03
    $101.03
    Each
       $101.03
    Supplier Number: 200-060-12
    Specifications:
    New Product
    New Product
  • Standard octagonal body vial with flat interior; Pressure rating: 60 PSI; Compatible with 21 mm trays
    3 ml
    17.3×26.4 mm
    PFA
    Non
    23 mm
    Log in for availability
    Log in for availability
    Each
    Each - $17.24
    $17.24
    Each
       $17.24
    Supplier Number: 200-003-12
    Specifications:
    New Product
    New Product
  • Standard octagonal body vial with conical interior; Pressure rating: 60 PSI; Compatible with 21 mm trays
    3 ml
    17.3×26.4 mm
    PFA
    Non
    23 mm
    Log in for availability
    Log in for availability
    Each
    Each - $19.34
    $19.34
    Each
       $19.34
    Supplier Number: 200-003-30
    Specifications:
    New Product
    New Product
  • Standard vial with conical interior, fin legs exterior; Pressure rating: 25 PSI; Compatible with 21 mm trays
    5 ml
    21.7×36.1 mm
    PFA
    Non
    24 mm
    Log in for availability
    Log in for availability
    Each
    Each - $22.42
    $22.42
    Each
       $22.42
    Supplier Number: 200-005-32
    Specifications:
    New Product
    New Product
  • Standard octagonal body vial with rounded interior; Pressure rating: 60 PSI; Compatible with 21 mm trays
    6 ml
    17.8×46.5 mm
    PFA
    Non
    24 mm
    Log in for availability
    Log in for availability
    Each
    Each - $29.63
    $29.63
    Each
       $29.63
    Supplier Number: 200-006-20
    Specifications:
    New Product
    New Product
  • Standard vial with flat interior; Pressure rating: 25 PSI; Compatible with 21 mm trays
    7 ml
    22.1×37.1 mm
    PFA
    Non
    24 mm
    Log in for availability
    Log in for availability
    Each
    Each - $20.57
    $20.57
    Each
       $20.57
    Supplier Number: 200-007-10
    Specifications:
    New Product
    New Product
  • Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    15 ml
    30.5×41.1 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $36.15
    $36.15
    Each
       $36.15
    Supplier Number: 200--015-12
    Specifications:
    New Product
    New Product
  • Standard vial with rounded interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    60 ml
    30.7×132.1 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $112.46
    $112.46
    Each
       $112.46
    Supplier Number: 200-060-20
    Specifications:
    New Product
    New Product
  • Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays
    60 ml
    30.7×132.1 mm
    PFA
    Non
    33 mm
    Log in for availability
    Log in for availability
    Each
    Each - $121.08
    $121.08
    Each
       $121.08
    Supplier Number: 200-060-30
    Specifications:
    New Product
    New Product
Accessories
Product Information
  • Catalog #
    Thickness
    Availability
    Price
  • PTFE/silicone laminates, ext. Ø 33 mm
    PTFE/silicone laminates, ext. Ø 33 mm
    78473-994
    PTFE: 0.25 mm; silicone: 2.3 mm
    Log in for availability
    Each - $10.99
Product Information
  • Catalog #
    Thickness
    Availability
    Price
  • PTFE/silicone laminates, ext. Ø 24 mm
    PTFE/silicone laminates, ext. Ø 24 mm
    78474-219
    PTFE: 0.25 mm; silicone: 2.3 mm
    Log in for availability
    Each - $10.08
Product Information
  • Catalog #
    Thickness
    Availability
    Price
  • PTFE/silicone laminates, ext. Ø 9 mm
    PTFE/silicone laminates, ext. Ø 9 mm
    78473-570
    PTFE: 0.25 mm; silicone: 1.3 mm
    Log in for availability
    Each - $9.54