Standard Vials
About this item
For 40 years, Savillex PFA vials have been the best choice for demanding trace metal applications, and are used in laboratories worldwide for sample preparation and storage.
Used typically for storage and also digestion in geochemistry
Flat base for maximum heating efficiency
Select the interior shape to suit the application
Square exterior vials with hex closures have higher pressure rating
Fin exterior vial used for microdistillation
Maximum service temperature of PFA is 260 ºC/500 ºF
The bottom type and high service temperature range makes them ideal for use directly on a hot plate. Molded from high purity virgin PFA, they are chemically inert, with ultra low trace metal content.
13 Options Available Below
Product Details & Documents
For 40 years, Savillex PFA vials have been the best choice for demanding trace metal applications, and are used in laboratories worldwide for sample preparation and storage.
Used typically for storage and also digestion in geochemistry
Flat base for maximum heating efficiency
Select the interior shape to suit the application
Square exterior vials with hex closures have higher pressure rating
Fin exterior vial used for microdistillation
Maximum service temperature of PFA is 260 ºC/500 ºF
The bottom type and high service temperature range makes them ideal for use directly on a hot plate. Molded from high purity virgin PFA, they are chemically inert, with ultra low trace metal content.
Product Family Options
Product Information
- DésignationCapacitéDiameter ×HMatièreAvec bouchonTaille du bouchonAvailabilityPrice
- Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays22 ml30.5×55.6 mmPFANon33 mmLog in for availabilityEach$43.26Supplier Number: 200-022-12Specifications:New ProductNew Product
- Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays22 ml30.5×55.6 mmPFANon33 mmLog in for availabilityEach$46.86Supplier Number: 200-022-30Specifications:New ProductNew Product
- Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays30 ml30.5×69.9 mmPFANon33 mmLog in for availabilityEach$58.55Supplier Number: 200-030-12Specifications:New ProductNew Product
- Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays30 ml30.5×79.0 mmPFANon33 mmLog in for availabilityEach$52.41Supplier Number: 200-030-30Specifications:New ProductNew Product
- Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays60 ml30.7×133.9 mmPFANon33 mmLog in for availabilityEach$101.03Supplier Number: 200-060-12Specifications:New ProductNew Product
- Standard octagonal body vial with flat interior; Pressure rating: 60 PSI; Compatible with 21 mm trays3 ml17.3×26.4 mmPFANon23 mmLog in for availabilityEach$17.24Supplier Number: 200-003-12Specifications:New ProductNew Product
- Standard octagonal body vial with conical interior; Pressure rating: 60 PSI; Compatible with 21 mm trays3 ml17.3×26.4 mmPFANon23 mmLog in for availabilityEach$19.34Supplier Number: 200-003-30Specifications:New ProductNew Product
- Standard vial with conical interior, fin legs exterior; Pressure rating: 25 PSI; Compatible with 21 mm trays5 ml21.7×36.1 mmPFANon24 mmLog in for availabilityEach$22.42Supplier Number: 200-005-32Specifications:New ProductNew Product
- Standard octagonal body vial with rounded interior; Pressure rating: 60 PSI; Compatible with 21 mm trays6 ml17.8×46.5 mmPFANon24 mmLog in for availabilityEach$29.63Supplier Number: 200-006-20Specifications:New ProductNew Product
- Standard vial with flat interior; Pressure rating: 25 PSI; Compatible with 21 mm trays7 ml22.1×37.1 mmPFANon24 mmLog in for availabilityEach$20.57Supplier Number: 200-007-10Specifications:New ProductNew Product
- Standard vial with flat interior; Pressure rating: 10 PSI; Compatible with 29 mm trays15 ml30.5×41.1 mmPFANon33 mmLog in for availabilityEach$36.15Supplier Number: 200--015-12Specifications:New ProductNew Product
- Standard vial with rounded interior; Pressure rating: 10 PSI; Compatible with 29 mm trays60 ml30.7×132.1 mmPFANon33 mmLog in for availabilityEach$112.46Supplier Number: 200-060-20Specifications:New ProductNew Product
- Standard vial with conical interior; Pressure rating: 10 PSI; Compatible with 29 mm trays60 ml30.7×132.1 mmPFANon33 mmLog in for availabilityEach$121.08Supplier Number: 200-060-30Specifications:New ProductNew Product
Accessories
Product Information
- Catalog #ThicknessAvailabilityPrice
- PTFE/silicone laminates, ext. Ø 33 mmPTFE/silicone laminates, ext. Ø 33 mm78473-994PTFE: 0.25 mm; silicone: 2.3 mmLog in for availabilityEach - $10.99
Product Information
- Catalog #ThicknessAvailabilityPrice
- PTFE/silicone laminates, ext. Ø 24 mmPTFE/silicone laminates, ext. Ø 24 mm78474-219PTFE: 0.25 mm; silicone: 2.3 mmLog in for availabilityEach - $10.08
Product Information
- Catalog #ThicknessAvailabilityPrice
- PTFE/silicone laminates, ext. Ø 9 mmPTFE/silicone laminates, ext. Ø 9 mm78473-570PTFE: 0.25 mm; silicone: 1.3 mmLog in for availabilityEach - $9.54