Vent Nut Assembly
¼" Vent nut assembly, PTFE
Catalog # 78473-744
New Product
Supplier: SAVILLEX
Fournisseur #: 550-740-300731
Jump to:
Product Details & Documents
Used to vent the vial cleaning system during heating. The adjustable vent nut assembly also enables smooth controlled liquid flow through the pour spout. Recommended annual replacement for vial cleaning system.
¹/₄" vent nut
9 mm PTFE filter membrane
PTFE retaining ring
Delivery information: Two years warranty.
Spécifications
- Désignation:¼" Vent nut assembly
- Matière:PTFE