Sealing Ring
Sealing ring, 40.1H×184.9Ø mm
78474-021
New Product
: SAVILLEX
: 550-740-400823
Fits cleaning container and silicone grip mat (78473-766) is recommended for tightening and loosening the sealing ring.
Upper service temperature of ECTFE is 150 °C (300 °F)
- Désignation:Sealing ring
- Dimensions:40.1H×184.9Ø mm