Vial Rack Systems | Avantor
Vial Rack Systems
New Product
Supplier:   SAVILLEX
Vial Rack Systems
Supplier:  SAVILLEX
New Product
Certificates
Vial Rack Systems
7 Options Available Below
Product Details & Documents
The Savillex vial rack system is the safest and most effi cient way to clean PFA vials, closures and other types of labware by acid soaking.
  • Comprises of a series of grid-like racks which stack on a center post
  • Racks are molded from PFA, excellent resistance to all mineral acids
  • Drip tray is included which can be used to stand the rack system on or to transfer it to a sink for rinsing
The vial rack system, holds vials and closures upright during cleaning, eliminating air pockets and also facilitates the loading and unloading of the vial cleaning system. In combination, the vial rack system and vial cleaning system provides a complete solution for the acid cleaning of Savillex PFA vials, closures and many other types of labware.
Three pre-confi gured vial rack systems are available, to accommodate the most popular sizes of Savillex vials: 7, 15 and 30 ml, along with appropriate closures. For vials or parts other than those listed above, a universal rack system is available. The universal rack system is designed to be used with any size vial or other items, therefore universal racks do not feature integrated spacers.
The rack system can also be used with most 4 L glass beakers. Since the rack system is too tall to allow the use of a watch glass, a purpose designed PFA beaker lid is available for when the system is used with a glass beaker.
Delivery information: Includes five PFA universal vial racks. Vial rack retainer included. Drip tray included.
Product Family Options
Individual vial racks
Informations sur le produit
  • Désignation
    Availability
    Price
  • 30 ml vial rack hold 12 Savillex 30 ml PFA vials, Height: 70.8 mm (2.79"); Int.Ø: 25.4 mm (1"); Ext.Ø: 152.4 mm (6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 159,50 $
    159,50 $
    Each
       159,50 $
    Supplier Number: 550-740-400891
    Spécifications:
    New Product
    New Product
  • 15 ml vial rack hold 12 Savillex 15 ml PFA Vials, Height: 41.75 mm (1.64"); Int.Ø: 25.4 mm (1"); Ext.Ø: 152.4 mm (6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 143,98 $
    143,98 $
    Each
       143,98 $
    Supplier Number: 550-740-400892
    Spécifications:
    New Product
    New Product
  • 7 ml vial rack hold 24 Savillex 7 ml PFA Vials, Height: 38.8 mm (1.53"); Int.Ø: 25.4 mm (1"); Ext.Ø: 152.4 mm (6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 159,50 $
    159,50 $
    Each
       159,50 $
    Supplier Number: 550-740-400893
    Spécifications:
    New Product
    New Product
Informations sur le produit
  • Désignation
    Availability
    Price
  • Vial rack system, 7 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 584,19 $
    1 584,19 $
    Each
       1 584,19 $
    Supplier Number: 550-07
    Spécifications:
    New Product
    New Product
  • Vial rack system, 15 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 451,06 $
    1 451,06 $
    Each
       1 451,06 $
    Supplier Number: 550-15
    Spécifications:
    New Product
    New Product
  • Vial rack system, 30 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 224,12 $
    1 224,12 $
    Each
       1 224,12 $
    Supplier Number: 550-30
    Spécifications:
    New Product
    New Product
  • Vial rack system, universal
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 893,52 $
    893,52 $
    Each
       893,52 $
    Supplier Number: 550-99
    Spécifications:
    New Product
    New Product
amed Group
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • PFA Beaker lid for vial rack system, Ø 165.6 mm (6.52")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 469,62 $
    469,62 $
    Each
       469,62 $
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.