Advanced Benchtop pH/ORP Meter
Product Details & Documents
High-precision benchtop pH and ORP meter with advanced data logging and diagnostics.
- Provides laboratory-grade pH and ORP measurement accuracy
- Supports multiple calibration points for precise standardization
- Built-in electrode diagnostics and system verification
- Automatic temperature compensation for stable results
- Large, color display with guided calibration and data visualization
- USB data export and GLP-compliant recordkeeping
- Ideal for research, quality control, and advanced laboratory coursework
The Advanced Benchtop pH/ORP Meter delivers professional-level accuracy and control for electrochemical analysis in laboratory and educational environments. Designed for research, quality assurance, and advanced instruction, this instrument provides high-resolution pH, oxidation-reduction potential (ORP), and temperature measurements with outstanding stability and repeatability. The system supports multi-point calibration with automatic buffer recognition and offers electrode diagnostics that alert users to aging or contamination, ensuring ongoing measurement reliability. Its automatic temperature compensation corrects readings for thermal variation, enabling consistent accuracy across diverse sample types and testing conditions.
A large, high-contrast color display provides clear visualization of readings, calibration data, and electrode performance, while an intuitive interface simplifies setup and daily operation. Built-in data logging and GLP compliance tools allow traceable recordkeeping for research and teaching laboratories. Designed for convenience and long-term reliability, the meter connects easily to a computer via USB for data transfer and backup, and it operates with a standard power adapter for continuous use in laboratory settings. The rugged benchtop design and advanced functionality make it a versatile choice for courses or facilities requiring dependable, research-quality electrochemical measurement.
Delivery information: Power Supply: Universal AC adapter (included).
Spécifications
- Désignation:Advanced benchtop pH/ORP meter
- Cat. No.:CA470360-148
- pH accuracy:±0.002 pH
- mV accuracy:±0.2 mV