Advanced Benchtop pH/ORP Meter | Avantor
Advanced Benchtop pH/ORP Meter
Advanced benchtop pH/ORP meter
Catalog # CA470360-148
New Product
Supplier:   Hanna
Fournisseur #:  HI6221
Advanced Benchtop pH/ORP Meter
Catalog # CA470360-148
Supplier:  Hanna
New Product
Advanced Benchtop pH/ORP Meter
Advanced Benchtop pH/ORP Meter
Advanced benchtop pH/ORP meter
Price
$1,504.88
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
High-precision benchtop pH and ORP meter with advanced data logging and diagnostics.
  • Provides laboratory-grade pH and ORP measurement accuracy
  • Supports multiple calibration points for precise standardization
  • Built-in electrode diagnostics and system verification
  • Automatic temperature compensation for stable results
  • Large, color display with guided calibration and data visualization
  • USB data export and GLP-compliant recordkeeping
  • Ideal for research, quality control, and advanced laboratory coursework
The Advanced Benchtop pH/ORP Meter delivers professional-level accuracy and control for electrochemical analysis in laboratory and educational environments. Designed for research, quality assurance, and advanced instruction, this instrument provides high-resolution pH, oxidation-reduction potential (ORP), and temperature measurements with outstanding stability and repeatability. The system supports multi-point calibration with automatic buffer recognition and offers electrode diagnostics that alert users to aging or contamination, ensuring ongoing measurement reliability. Its automatic temperature compensation corrects readings for thermal variation, enabling consistent accuracy across diverse sample types and testing conditions.
A large, high-contrast color display provides clear visualization of readings, calibration data, and electrode performance, while an intuitive interface simplifies setup and daily operation. Built-in data logging and GLP compliance tools allow traceable recordkeeping for research and teaching laboratories. Designed for convenience and long-term reliability, the meter connects easily to a computer via USB for data transfer and backup, and it operates with a standard power adapter for continuous use in laboratory settings. The rugged benchtop design and advanced functionality make it a versatile choice for courses or facilities requiring dependable, research-quality electrochemical measurement.
Delivery information: Power Supply: Universal AC adapter (included).
Spécifications
  • Désignation:
    Advanced benchtop pH/ORP meter
  • Cat. No.:
    CA470360-148
  • pH accuracy:
    ±0.002 pH
  • mV accuracy:
    ±0.2 mV
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.