Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
edge® Multiparameter pH Meter

edge® Multiparameter pH Meter

edge® Multiparameter pH meter
Catalog # CA470360-146
New Product
Supplier:   Hanna
Supplier #:  HI2020
edge® Multiparameter pH Meter
Catalog # CA470360-146
Supplier:  Hanna
New Product
edge® Multiparameter pH Meter
edge® Multiparameter pH Meter

edge® Multiparameter pH meter

Price
$826.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Versatile digital meter for pH, conductivity, and dissolved oxygen measurement in one device.

  • Measures pH, conductivity, total dissolved solids (TDS), salinity, and dissolved oxygen
  • Large backlit LCD with wide viewing angle for classroom or lab visibility
  • Smart electrode recognition automatically loads calibration and sensor data
  • Capacitive touch keypad for smooth, sealed operation
  • Can be used as a benchtop, wall-mounted, or portable meter
  • Automatic calibration and temperature compensation ensure precision
  • USB port for easy data transfer and record keeping

The edge® Multiparameter pH Meter combines advanced measurement capabilities with a modern, lightweight design ideal for laboratory, classroom, or field environments. This digital instrument allows quick, accurate analysis of pH, conductivity, total dissolved solids (TDS), salinity, and dissolved oxygen - all within a single device. The meter recognizes interchangeable smart electrodes automatically, retrieving stored calibration information to simplify setup and prevent user error. Automatic temperature compensation provides consistent readings across diverse environmental and sample conditions. With its slim, tablet-style design, this instrument can function as a benchtop unit, wall-mounted system, or portable field meter, adapting easily to space and workflow requirements. The large, high-contrast display and responsive capacitive keypad allow intuitive operation and clear data presentation, even in bright laboratory lighting.

Data can be easily stored or transferred via USB, supporting both classroom data logging and professional reporting. Rechargeable battery operation ensures long-lasting performance for field or bench use. Engineered for durability and flexibility, the edge® Multiparameter Meter supports high-accuracy measurements while remaining accessible for students learning water quality testing, chemistry, or environmental analysis.

Delivery information: Rechargeable battery and 5 V DC adapter (included).

Specifications
  • Désignation:
    edge® Multiparameter pH meter
  • Cat. No.:
    CA470360-146
  • pH accuracy:
    ±0.01 pH
  • Dimensions:
    202×140×12.7 mm (8×5.5×0.5")
  • Weight:
    250 g (8.8 oz.)

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.