pH 10 Buffer Solution
pH 10 buffer solution, 500 ml bottle
  CA470357-990
 :   Hanna
 :  HI7010L
pH 10 Buffer Solution
  CA470357-990
 :  Hanna
 :  
pH 10 Buffer Solution
pH 10 Buffer Solution
pH 10 buffer solution, 500 ml bottle
 :  
$90.38
Each
High-quality pH 10 calibration buffer for standardizing meters in basic solutions.
  • Provides accurate pH 10 calibration for instruments used in alkaline samples
  • Traceable to NIST standards for reliable performance
  • Clearly labeled with lot number and expiration date for quality assurance
  • Airtight, tamper-proof bottle maintains solution integrity
  • Long shelf life ensures dependable classroom or laboratory use
  • Ideal for calibrating pH meters used in environmental and chemistry labs
The pH 10 Buffer Solution (500 mL) provides a dependable reference for calibrating pH meters in basic or alkaline conditions. This standard solution is traceable to NIST standards, ensuring precise and consistent results across laboratory, classroom, and field applications. Each bottle includes a lot number and expiration date clearly printed for traceability and quality control. The solution is packaged in an airtight, tamper-evident HDPE bottle to prevent contamination and evaporation, maintaining stability over extended storage periods.
Formulated for long-term reliability, this buffer remains stable for up to five years when unopened, making it an excellent choice for teaching laboratories and research facilities that require consistent calibration accuracy. Ideal for use in chemistry, environmental science, and general laboratory experiments, this buffer allows students and professionals to standardize their instruments confidently when measuring alkaline samples such as detergents, water treatment solutions, or biological media.
  • Désignation:
    pH 10 buffer solution, 500 ml bottle