Masterflex® L/S® Variable Speed Analog Console Pump Systems
Masterflex® L/S® Variable Speed Analog Console Pump Systems
New Product
Masterflex® L/S® Variable Speed Analog Console Pump Systems
New Product
Masterflex® L/S® Variable Speed Analog Console Pump Systems
Don’t lose control of your metering applications - digital display gives you precise control.
  • ABS plastic and aluminum housing
  • Accurate and repeatable settings with three-digit LED display of rpm
  • Pump fluid in either direction with reversible motor - perfect for purging tubing
  • Choose multichannel systems for synchronous flow from all channels
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP 33-rated drives
  • Two ways to control speed, direction, and start/stop - use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Certifications: CE Compliant.
  • Model:
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • 77926-20
  • 77917-00
  • 77917-10
  • 77917-20
  • 77917-30
  • No. of channels:
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • 4
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • 4
  • 1
  • 1
  • 1
  • 1
  • No. of rollers:
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • 3
  • 3
  • 3
  • 3
  • 6
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • 6
  • 3
  • 3
  • 3
  • 3
  • Control type:
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Variable-Speed Digital
  • Housing material:
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • ABS plastic and aluminium
  • Min. flow rate per channel:
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • Max. flow rate per channel:
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 47 ml/min
  • 480 ml/min
  • 1700 ml/min
  • 480 ml/min
  • 1700 ml/min
  • Min. flow rate:
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • 0.47 ml/min
  • 4.8 ml/min
  • 17 ml/min
  • Drive IP rating:
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • IP 33
  • Accuracy:
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
  • Anti-drip:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Cloud connected:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Digital dispensing:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Drive:
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • 07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
  • Duty cycle:
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Continuous
  • Min. RPM:
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 1 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • Open-head sensor:
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • No
  • No
  • Yes
  • Yes
  • Power:
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • 90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Remote I/O:
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Tubing:
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16 tube set, pk of 8
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Masterflex® L/S® 16, 10'
  • Masterflex® L/S® 24, 10'
  • Tubing sizes accepted:
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® two-stop tube sets: L/S® 13, 14, 16
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • L/S® 13, 14, 16, 25, 17, 18
  • L/S® 15, 24, 35, 36
  • Washdown capable:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Current:
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • 1.8 A @ 115 V; 1.2 A @ 230 V
  • Frequency:
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • Voltage:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Product Family Options
  • Désignation
    Plug Type
    Débit maximal
    Hauteur
    Longueur
    Width
    Pump Head
    Max. RPM
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    IT
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-IT
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    IT
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-IT
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IT
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-IT
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    IT
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-IT
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IT
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-IT
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    IL
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-IL
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    IL
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-IL
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IL
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-IL
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    IL
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-IL
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IL
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-IL
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    CH
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-CH
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    CH
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-CH
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    CH
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-CH
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    CH
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-CH
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    CH
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-CH
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    AU
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-AU
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    AU
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-AU
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    AU
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-AU
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    AU
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-AU
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    AU
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-AU
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    DK
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-DK
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    DK
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-DK
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    DK
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-DK
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    DK
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-DK
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    DK
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-DK
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    IN
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-IN
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    IN
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-IN
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IN
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-IN
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    IN
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-IN
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    IN
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-IN
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    EU
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-EU
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    EU
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-EU
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    EU
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-EU
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    EU
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-EU
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    EU
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-EU
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    5000
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20-UK
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    5000
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00-UK
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    5000
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10-UK
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    5000
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20-UK
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    5000
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30-UK
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Multichannel Pump Head for Precision Tubing, 4-Channel; 90 - 260 VAC
    US
    188 ml/min
    215.9 mm (8.5")
    349 mm (13.74")
    210.8 mm (8.3")
    07535-04 (L/S® 4-channel pump head)
    100 rpm
    Each
    Each - $5,510.00
    $5,510.00
    Each
       $5,510.00
    New Product
    Specifications
    Cat. No.
    MFLX77926-20
    Model
    77926-20
    No. of channels
    4
    No. of rollers
    6
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    0.47 ml/min
    Max. flow rate per channel
    47 ml/min
    Min. flow rate
    0.47 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-30 (L/S® Variable-Speed Drive with Remote I/O, 100 rpm)
    Duty cycle
    Continuous
    Min. RPM
    1 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16 tube set, pk of 8
    Tubing sizes accepted
    L/S® two-stop tube sets: L/S® 13, 14, 16
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II Pump Head for Precision Tubing; 90 - 260 VAC
    US
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-60 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-00
    Model
    77917-00
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S®Precision Variable-Speed Pump with Easy-Load® II Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    US
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-62 (L/S® Easy-Load® II pump head)
    600 rpm
    Each
    Each - $4,440.00
    $4,440.00
    Each
       $4,440.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-10
    Model
    77917-10
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    No
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for Precision Tubing; 90 - 260 VAC
    US
    480 ml/min
    215.9 mm (8.5")
    352.6 mm (13.88")
    210.8 mm (8.3")
    77200-30 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-20
    Model
    77917-20
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    4.8 ml/min
    Max. flow rate per channel
    480 ml/min
    Min. flow rate
    4.8 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 16, 10'
    Tubing sizes accepted
    L/S® 13, 14, 16, 25, 17, 18
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product
  • L/S® Precision Variable-Speed Pump with Easy-Load® II OHS Pump Head for High-Performance Precision Tubing; 90 - 260 VAC
    US
    1700 ml/min
    215.9 mm (8.5")
    339.9 mm (13.38")
    210.8 mm (8.3")
    77200-32 (L/S® Easy-Load® II OHS pump head)
    600 rpm
    Each
    Each - $4,650.00
    $4,650.00
    Each
       $4,650.00
    New Product
    Specifications
    Cat. No.
    MFLX77917-30
    Model
    77917-30
    No. of channels
    1
    No. of rollers
    3
    Control type
    Variable-Speed Digital
    Housing material
    ABS plastic and aluminium
    Min. flow rate per channel
    17 ml/min
    Max. flow rate per channel
    1700 ml/min
    Min. flow rate
    17 ml/min
    Drive IP rating
    IP 33
    Accuracy
    ±0.25% (1 rpm at 600 and 300 rpm; 0.1 rpm at 100 rpm)
    Anti-drip
    No
    Cloud connected
    No
    Digital dispensing
    No
    Drive
    07527-10 (L/S® Variable-Speed Drive with Remote I/O, 600 rpm)
    Duty cycle
    Continuous
    Min. RPM
    6 rpm
    Open-head sensor
    Yes
    Power
    90 - 260 VAC; 50/60 Hz; 1.8 A @ 115 V; 1.2 A @ 230 V
    Pump series
    L/S
    Remote I/O
    4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure
    Reversible motor
    Yes
    Tubing
    Masterflex® L/S® 24, 10'
    Tubing sizes accepted
    L/S® 15, 24, 35, 36
    Washdown capable
    No
    Current
    1.8 A @ 115 V; 1.2 A @ 230 V
    Frequency
    50/60 Hz
    Voltage
    90 - 260 VAC
    Warranty
    2 years
    New Product