Vasculitis PBMC
Vasculitis PBMC, Cryopreserved, Single donor, 5×10⁶ cells (Vial), Human, adult
78170-584
New Product
: VAS-PBMC-10M
Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed vasculitis donors for immunology and translational research.
Clinically confirmed vasculitis donors
High-viability cryopreservation
Supports flow, cytokine, and gene-expression assays
Documentation available
Vasculitis PBMC 5M are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in vasculitis.
- Format de cellule:Cryopreserved
- Donneur unique/donneurs regroupés:Single donor
- Désignation:Vasculitis PBMC
- Taille:5×10⁶ cells (Vial)
- Source:Human, adult



