COPD PBMC
New Product
COPD PBMC
New Product
COPD PBMC
Cryopreserved peripheral blood mononuclear cells (PBMCs) from clinically confirmed COPD donors for immunology and translational research.
  • Clinically confirmed COPD donors
    High-viability cryopreservation
    Research-ready aliquots
    Supports flow, cytokine, and gene-expression assays
    Documentation available
COPD PBMC are collected under IRB oversight and processed to preserve cellular integrity and functional responses. The PBMC format is suitable for phenotyping, cytokine release assays, co-culture systems, and multi-omics workflows investigating immune mechanisms in copd.
Product Family Options
  • Désignation
    Format de cellule
    Donneur unique/donneurs regroupés
    Taille
    Source
  • COPD PBMC
    Cryopreserved
    Single donor
    2×10⁹ cells (Vial, bulk)
    Human, adult
    Each
    Each - 21 100,65 $
    21 100,65 $
    Each
       21 100,65 $
     : CPD-LKLT-2B
    New Product
    New Product
  • COPD PBMC
    Cryopreserved
    Single donor
    1×10⁸ cells (Vial)
    Human, adult
    Each
    Each - 1 856,86 $
    1 856,86 $
    Each
       1 856,86 $
     : CPD-LKLT-100M
    New Product
    New Product
  • COPD PBMC
    Cryopreserved
    Single donor
    1×10⁷ cells (Vial)
    Human, adult
    Each
    Each - 835,59 $
    835,59 $
    Each
       835,59 $
     : CPD-LKLT-10M
    New Product
    New Product
  • COPD PBMC
    Cryopreserved
    Single donor
    2.5×10⁷ cells (Vial)
    Human, adult
    Each
    Each - 1 181,65 $
    1 181,65 $
    Each
       1 181,65 $
     : CPD-LKLT-25M
    New Product
    New Product
  • COPD PBMC
    Cryopreserved
    Single donor
    5×10⁷ cells (Vial)
    Human, adult
    Each
    Each - 1 671,18 $
    1 671,18 $
    Each
       1 671,18 $
     : CPD-LKLT-50M