SparQ Lysis Kits
À propos de cet article
High-quality, high molecular weight gDNA extraction from DBS for high-throughput workflows.
Rapid workflow: Extracts high quality, double-stranded gDNA in 50 minutes, saving time and increasing laboratory efficiency
- High molecular weight gDNA: Yields intact genomic DNA (>50 kb) ideal for long-read sequencing
- Versatile sample compatibility: One kit supports a broad range of sample types, including DBS, saliva, and cheek swab
- Broad application use: Delivers DNA ready for NGS (short and long-read), qPCR, dPCR, and other genomic workflows
- Automation-ready: Compatible with high-throughput and automated platforms for effortless scalability
The sparQ Lysis Kit offers a fast and efficient method for extracting high-molecular-weight genomic DNA (>50 kb) from a variety of sample types, including dried blood spots (DBS), whole blood, saliva, and cheek swab - in just 50 minutes. Designed for both manual and automated platforms, this streamlined workflow consistently delivers high gDNA yield results and is optimized for high-throughput processing.
The sparQ Lysis Kit is a key component of the sparQ DBS-seq Workflow, an end-to-end solution for whole genome sequencing (WGS) library preparation from DBS. As the sample preparation module of this Workflow, the sparQ Lysis Kit enables efficient extraction of high-quality genomic DNA for next-generation sequencing, even from limited sample input. The complete Workflow also includes library preparation and library quantification, providing a streamlined and scalable solution.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.
Caution: sparQ Lysis kit is intended for molecular biology applications. This product is not intended for the diagnosis, prevention or treatment of a disease.
2 Options Available Below
Product Details & Documents
High-quality, high molecular weight gDNA extraction from DBS for high-throughput workflows.
Rapid workflow: Extracts high quality, double-stranded gDNA in 50 minutes, saving time and increasing laboratory efficiency
- High molecular weight gDNA: Yields intact genomic DNA (>50 kb) ideal for long-read sequencing
- Versatile sample compatibility: One kit supports a broad range of sample types, including DBS, saliva, and cheek swab
- Broad application use: Delivers DNA ready for NGS (short and long-read), qPCR, dPCR, and other genomic workflows
- Automation-ready: Compatible with high-throughput and automated platforms for effortless scalability
The sparQ Lysis Kit offers a fast and efficient method for extracting high-molecular-weight genomic DNA (>50 kb) from a variety of sample types, including dried blood spots (DBS), whole blood, saliva, and cheek swab - in just 50 minutes. Designed for both manual and automated platforms, this streamlined workflow consistently delivers high gDNA yield results and is optimized for high-throughput processing.
The sparQ Lysis Kit is a key component of the sparQ DBS-seq Workflow, an end-to-end solution for whole genome sequencing (WGS) library preparation from DBS. As the sample preparation module of this Workflow, the sparQ Lysis Kit enables efficient extraction of high-quality genomic DNA for next-generation sequencing, even from limited sample input. The complete Workflow also includes library preparation and library quantification, providing a streamlined and scalable solution.
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.
Caution: sparQ Lysis kit is intended for molecular biology applications. This product is not intended for the diagnosis, prevention or treatment of a disease.
Product Family Options
Informations sur le produit
- DésignationNo. of ReactionsAvailabilityPrice
- Supplier Number: 95220-024Spécifications:New ProductNew Product
- Supplier Number: 95220-096Spécifications:New ProductNew Product