High Capacity Racking Systems | Avantor
High Capacity Racking Systems
High Capacity Racking Systems
Certificates
High Capacity Racking Systems
High Capacity Racking Systems
14 Options Available Below
Product Details & Documents
Our racking systems are precision-engineered to provide maximum storage capacity, organization, and durability inside laboratory and isothermal freezers.
  • Choice of materials
  • Stainless steel: Provides long-term corrosion resistance, strength, and durability under repeated freeze-thaw cycles. Ideal for high-demand research or GMP environments
  • Aluminum: Lightweight yet robust for easy handling and installation, offering excellent thermal conductivity and reduced mass for fast temperature recovery
  • Lifetime warranty on workmanship
Designed to fit a variety of freezer diameters - including 20, 21, 31 and 39-inch isothermal freezer models - these systems are ideal for laboratories, biobanks, and research facilities requiring efficient cryogenic sample management.
Constructed from stainless steel or aluminum, each system delivers superior corrosion resistance and structural integrity under ultra-low and cryogenic temperature conditions. Whether used in vapor-phase liquid nitrogen (LN₂) storage or mechanical freezer environments, these racks maintain consistent performance and easy accessibility during long-term sample storage.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Aluminum 7-rack system for 21" diameter freezer (V- 1500 series only)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 765,38 $
    1 765,38 $
    Each
       1 765,38 $
    Supplier Number: 2001A-C100S
  • Stainless steel 7-rack system for 20 or 21" diameter freezer (V-1500 or S-1500 series)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 042,10 $
    2 042,10 $
    Each
       2 042,10 $
    Supplier Number: 2001S-C100
  • Stainless steel 7-rack system for 21" diameter freezer (V-1500 series only)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 179,35 $
    2 179,35 $
    Each
       2 179,35 $
    Supplier Number: 2001S-C100S
  • Stainless steel 7-rack system for 20 or 21" diameter freezer (V-1500 or S-1500 series)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 027,08 $
    2 027,08 $
    Each
       2 027,08 $
    Supplier Number: 2001S-C81
  • Stainless steel 7-rack system for 20 or 21" diameter freezer (V-1500 or S-1500 series)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 564,42 $
    1 564,42 $
    Each
       1 564,42 $
    Supplier Number: 2007S-C100
  • Vertical rack system for V-1500AB. 20 stainless steel racks, 8100 vial capacity
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 185,97 $
    2 185,97 $
    Each
       2 185,97 $
    Supplier Number: 21-1209V-C81
  • Aluminum 17-rack system for 31" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 4 287,34 $
    4 287,34 $
    Each
       4 287,34 $
    Supplier Number: 3101A-C100S
  • Stainless steel 17-rack system for 31" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 3 724,17 $
    3 724,17 $
    Each
       3 724,17 $
    Supplier Number: 3101S-C100
  • Stainless steel 17-rack system for 31" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 3 724,17 $
    3 724,17 $
    Each
       3 724,17 $
    Supplier Number: 3101S-C100S
  • Stainless steel 17-rack system for 31" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 3 724,17 $
    3 724,17 $
    Each
       3 724,17 $
    Supplier Number: 3101S-C81
  • Aluminum 28-rack system for 39" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 7 061,59 $
    7 061,59 $
    Each
       7 061,59 $
    Supplier Number: 3301A-C100S
  • Stainless steel 28-rack system for 39" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 6 133,95 $
    6 133,95 $
    Each
       6 133,95 $
    Supplier Number: 3301S-C100S
  • Aluminum 28-rack system for 39" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 8 936,21 $
    8 936,21 $
    Each
       8 936,21 $
    Supplier Number: 3325A-C100S
  • Stainless steel 28-rack system for 39" diameter freezer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 7 681,64 $
    7 681,64 $
    Each
       7 681,64 $
    Supplier Number: 3325S-C100S