Multi-Use Press for 48 Filter Vials
Multi-use press for 48 filter vials, metal, non sterile
76808-506
: 35015
The Thomson multi-use press quickly depresses autosampler-ready filter vials.
Presses up to 48 autosampler ready filter vials at a time
- Works with 48 position block; block fits some autosamplers
- Easily automate filter vial pressing
- Works with all Thomson filter vials
The 48 position block can be transferred to a robot for automation.
- Désignation:Multi-use press for 48 filter vials, metal, non sterile