Agar Bacteriological, qualité Bactériologie
About this item
A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.
Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.
- CAS Number:9002-18-0
- Formula:(C₁₂H₁₈O₉)n
2 Options Available Below
Product Details & Documents
A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.
Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.
- CAS Number:9002-18-0
- Formula:(C₁₂H₁₈O₉)n
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



