Filter Holder, Swinney | Avantor
Filter Holder, Swinney
13 mm filter holder, acetal copo, swinney
Catalog # 78172-644
New Product
Fournisseur #:  1220950
Filter Holder, Swinney
Catalog # 78172-644
New Product
Filter Holder, Swinney
13 mm filter holder, acetal copo, swinney
Price
$746.20
Log in pour la disponibilité
Pack of 5
Product Details & Documents
The Swinney 13 mm filter holder is optimized for small volume (1 - 5 ml) particulate removal from fluids dispensed with a syringe.
  • Highly resistant to organic components No need for specific tools Quick, efficient assembly
The holder is resistant to alcohols, esters, ethers, glycols, aromatic hydrocarbons, halogenated hydrocarbons, ketones, oils, photoresists, and many other chemicals. Although suitable for most weak acids and bases, acid compatibility tests are recommended.
Spécifications
  • Désignation:
    13 mm filter holder, acetal copo, swinney