Microbial Count Plate
New Product
Microbial Count Plate
New Product
Microbial Count Plate
GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.
  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required
GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.
Product Family Options
  • Désignation
    Format
  • Pivot E. coli/Coliform count plate
    Dark purple red colony, 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $349.35
    $349.35
    Pack of 25
       $349.35
     : MCPG00325
    New Product
    New Product
  • Shigella count plate
    Red colony, , 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG02525
    New Product
    New Product
  • Bacillus Psychrophilus count plate
    Black, green colony, 36±1 °C, 26±2h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG01925
    New Product
    New Product
  • Aerobic bacillus count plate
    Red colony, 21±1 °C, 48-60h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG02425
    New Product
    New Product
  • Enterobacteriaceae EB count plate
    Purple, red colony, 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $194.08
    $194.08
    Pack of 25
       $194.08
     : MCPG01625
    New Product
    New Product
  • Listeria count plate
    Green colony, 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $232.89
    $232.89
    Pack of 25
       $232.89
     : MCPG00525
    New Product
    New Product
  • Coliform count plate
    Green colony, 28±1 °C, 48-2h incubation
    Pack of 25
    Pack of 25 - $97.05
    $97.05
    Pack of 25
       $97.05
     : MCPG01025
    New Product
    New Product
  • Lactic Acid Bacteria count plate
    Purple, red colony, 36±1 °C, 48±2h incubation
    Pack of 25
    Pack of 25 - $465.80
    $465.80
    Pack of 25
       $465.80
     : MCPG01425
    New Product
    New Product
  • Enterococcus faecalis count plate
    Red colony, 55±1 °C, 36±1h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG01825
    New Product
    New Product
  • Salmonella count plate
    Red colony, 36±1 °C, 48±2h incubation
    Pack of 25
    Pack of 25 - $194.08
    $194.08
    Pack of 25
       $194.08
     : MCPG01525
    New Product
    New Product
  • Staph count plate
    Red colony, 36±1 °C, 48±2h incubation
    Pack of 25
    Pack of 25 - $135.86
    $135.86
    Pack of 25
       $135.86
     : MCPG00225
    New Product
    New Product
  • Yeast and molds platecount plate
    Blue green colony, 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $97.05
    $97.05
    Pack of 25
       $97.05
     : MCPG00825
    New Product
    New Product
  • Vibrio parahaemolyticus count plate
    Gray colony with halo, 36±1 °C, 18-24h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG02825
    New Product
    New Product
  • Escherichia coli O157 count plate
    Blue, green colony, 36±1 °C, 36h incubation
    Pack of 25
    Pack of 25 - $465.80
    $465.80
    Pack of 25
       $465.80
     : MCPG02725
    New Product
    New Product
  • Geobacillus stearothermophilus count plate
    Red colony, 36±1 °C, 18-24h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG01725
    New Product
    New Product
  • Pivot Coliform count plate
    Blue purple colony, 36±1 °C, 24±2h incubation
    Pack of 25
    Pack of 25 - $349.35
    $349.35
    Pack of 25
       $349.35
     : MCPG00425
    New Product
    New Product
  • Aerobic count plate
    Pack of 25
    Pack of 25 - $97.05
    $97.05
    Pack of 25
       $97.05
     : MCPG00125
    New Product
    New Product
  • Psychrophilic Bacteria count plate
    Red colony, 36±1 °C, 8-18h incubation
    Pack of 25
    Pack of 25 - $388.16
    $388.16
    Pack of 25
       $388.16
     : MCPG02925
    New Product
    New Product
  • E. coli/Coliform count plate
    Red colony, 36±1 °C, 18-24h incubation
    Pack of 25
    Pack of 25 - $97.05
    $97.05
    Pack of 25
       $97.05
     : MCPG01125
    New Product
    New Product
  • Bacillus Cereus count plate
    Bule purple red colony, 36±1 °C, 18-24h incubation
    Pack of 25
    Pack of 25 - $194.08
    $194.08
    Pack of 25
       $194.08
     : MCPG01325
    New Product
    New Product
  • Listeria monocytogenes count plate
    Red colony, 36 ±1 °C, 24-48h incubation
    Pack of 25
    Pack of 25 - $465.80
    $465.80
    Pack of 25
       $465.80
     : MCPG02625