Extraction Thimble Filter, Glass Microfiber
New Product
Extraction Thimble Filter, Glass Microfiber
New Product
Extraction Thimble Filter, Glass Microfiber
GVS extraction thimble filters, glass microfiber, available in multuple sizes.
  • Manufactured in high-alpha cellulose cotton linters Strong mechanical structure and retentivity Maximum working temperature: 120 °C
The extraction thimbles are suitable for Soxhlet type, Tecator type, or similar devices. They are located in the extractor body, used to accommodate a sample of solid material to extract certain components out, with the addition of an appropriate solvent.
Product Family Options
  • Taille
  • 43×123 mm
    Pack of 25
    Pack of 25 - $1,357.79
    $1,357.79
    Pack of 25
       $1,357.79
     : ET43123GF0Q00
    New Product
    New Product
  • 19×90 mm
    Pack of 25
    Pack of 25 - $722.68
    $722.68
    Pack of 25
       $722.68
     : ET19090GF0Q00
    New Product
    New Product
  • 35×150 mm
    Pack of 25
    Pack of 25 - $1,102.94
    $1,102.94
    Pack of 25
       $1,102.94
     : ET35150GF0Q00