Extraction Thimble Filter, Glass Microfiber
New Product
GVS extraction thimble filters, glass microfiber, available in multuple sizes.
- Manufactured in high-alpha cellulose cotton linters Strong mechanical structure and retentivity Maximum working temperature: 120 °C
The extraction thimbles are suitable for Soxhlet type, Tecator type, or similar devices. They are located in the extractor body, used to accommodate a sample of solid material to extract certain components out, with the addition of an appropriate solvent.
GVS extraction thimble filters, glass microfiber, available in multuple sizes.
- Manufactured in high-alpha cellulose cotton linters Strong mechanical structure and retentivity Maximum working temperature: 120 °C
The extraction thimbles are suitable for Soxhlet type, Tecator type, or similar devices. They are located in the extractor body, used to accommodate a sample of solid material to extract certain components out, with the addition of an appropriate solvent.
Product Family Options
- Taille
- : ET43123GF0Q00New ProductNew Product
- : ET19090GF0Q00New ProductNew Product
- : ET35150GF0Q00New ProductNew Product