Quartz Microfiber Filter, Pure
About this item
Quartz microfiber filter, D0QF2 pure grade, binder-free, with low trace levels of heavy metals.
- Excellent retention of very fine particles Exceptional chemical and thermal resistance Excellent weight and dimensional stability with lowest trace metal content High permeation enables large volume of air to pass through Higher temperature stability than glass microfiber filters; up to 900 °C Excellent chemical stability with practically no filter-mass loss in the presence of acid gases
GVS Quartz microfiber filters are made with 100% pure quartz microfiber with zero binders. Exhibit greater chemical resistance at high temperatures than glass microfiber. Excellent choice for use in environments with extreme temperature up to 900°C and/or aggressive chemical exposure. Retention loading and air flow permeation similar to glass microfiber filters. Use wherever filters of the highest purity are needed.
11 Options Available Below
Product Details & Documents
Quartz microfiber filter, D0QF2 pure grade, binder-free, with low trace levels of heavy metals.
- Excellent retention of very fine particles Exceptional chemical and thermal resistance Excellent weight and dimensional stability with lowest trace metal content High permeation enables large volume of air to pass through Higher temperature stability than glass microfiber filters; up to 900 °C Excellent chemical stability with practically no filter-mass loss in the presence of acid gases
GVS Quartz microfiber filters are made with 100% pure quartz microfiber with zero binders. Exhibit greater chemical resistance at high temperatures than glass microfiber. Excellent choice for use in environments with extreme temperature up to 900°C and/or aggressive chemical exposure. Retention loading and air flow permeation similar to glass microfiber filters. Use wherever filters of the highest purity are needed.
Product Family Options
Product Information
- GradeØÉpaisseurWeightAvailabilityPrice
- Supplier Number: FP110D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP090D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP050D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP150D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP125D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP055D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP070D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP120D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP037D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP025D0QF2QUFC01Specifications:New ProductNew Product
- Supplier Number: FP047D0QF2QUFC01Specifications:New ProductNew Product