Quartz Microfiber Filter, Pure
New Product
Quartz Microfiber Filter, Pure
New Product
Certificates
Quartz Microfiber Filter, Pure
11 Options Available Below
Product Details & Documents
Quartz microfiber filter, D0QF2 pure grade, binder-free, with low trace levels of heavy metals.
  • Excellent retention of very fine particles Exceptional chemical and thermal resistance Excellent weight and dimensional stability with lowest trace metal content High permeation enables large volume of air to pass through Higher temperature stability than glass microfiber filters; up to 900 °C Excellent chemical stability with practically no filter-mass loss in the presence of acid gases
GVS Quartz microfiber filters are made with 100% pure quartz microfiber with zero binders. Exhibit greater chemical resistance at high temperatures than glass microfiber. Excellent choice for use in environments with extreme temperature up to 900°C and/or aggressive chemical exposure. Retention loading and air flow permeation similar to glass microfiber filters. Use wherever filters of the highest purity are needed.
Product Family Options
Product Information
  • Grade
    Ø
    Épaisseur
    Weight
    Availability
    Price
  • Pure
    110 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $2,368.18
    $2,368.18
    Pack of 100
       $2,368.18
    Supplier Number: FP110D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    90 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,793.23
    $1,793.23
    Pack of 100
       $1,793.23
    Supplier Number: FP090D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    50 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $829.37
    $829.37
    Pack of 100
       $829.37
    Supplier Number: FP050D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    150 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $4,819.21
    $4,819.21
    Pack of 100
       $4,819.21
    Supplier Number: FP150D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    125 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $3,013.59
    $3,013.59
    Pack of 100
       $3,013.59
    Supplier Number: FP125D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    55 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $988.53
    $988.53
    Pack of 100
       $988.53
    Supplier Number: FP055D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    70 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,280.89
    $1,280.89
    Pack of 100
       $1,280.89
    Supplier Number: FP070D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    120 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $2,918.96
    $2,918.96
    Pack of 100
       $2,918.96
    Supplier Number: FP120D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    37 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $431.84
    $431.84
    Pack of 100
       $431.84
    Supplier Number: FP037D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    25 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $336.22
    $336.22
    Pack of 100
       $336.22
    Supplier Number: FP025D0QF2QUFC01
    Specifications:
    New Product
    New Product
  • Pure
    47 mm
    430 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $616.38
    $616.38
    Pack of 100
       $616.38
    Supplier Number: FP047D0QF2QUFC01
    Specifications:
    New Product
    New Product