Polyethersulfone (PES) Membranes | Avantor
Polyethersulfone (PES) Membranes
New Product
Polyethersulfone (PES) Membranes
New Product
Certificates
9 Options Available Below
Product Details & Documents
Polyethersulfone membranes are hydrophilic, with a wide range of pore sizes.
  • Hydrophilic format eliminates the need for wetting agents that can potentially interfere with analyses
  • Low extractables ensure test results will not be compromised by wetting agents or other extractables
  • Low drug and protein binding maximizes recovery of critical drugs or proteins
  • Superior burst strength protects the integrity of the membrane under high pressure
  • Quality checks, both down and across the membrane, ensure dependable results every time
Polyethersulfone (PES) Filtration Membrane is hydrophilic and cast from pure polyethersulfone polymer. It is designed to remove particulates during general filtration and its low protein and drug binding characteristics make it ideally suited for use in life science applications.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 78173-448
  • 78173-644
  • 78173-652
  • 78172-978
  • 78172-488
  • 78172-350
  • 78173-310
  • 78172-234
  • 78172-756
  • Pore size:
  • 0.22 μm
  • 0.45 μm
  • 1.2 μm
  • 0.45 μm
  • 0.1 μm
  • 0.1 μm
  • 0.22 μm
  • 0.22 μm
  • 1.2 μm
Spécifications
Product Family Options
Informations sur le produit
  • Taille de pore
    Ø
    Availability
    Price
  • 0.22 μm
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $43.00
    $43.00
    Pack of 100
       $43.00
    Supplier Number: 1214193
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78173-448
    Pore size
    0.22 μm
    New Product
  • 0.45 μm
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $43.54
    $43.54
    Pack of 100
       $43.54
    Supplier Number: 1214532
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78173-644
    Pore size
    0.45 μm
    New Product
  • 1.2 μm
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $50.90
    $50.90
    Pack of 100
       $50.90
    Supplier Number: 1222267
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78173-652
    Pore size
    1.2 μm
    New Product
  • 0.45 μm
    90 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $51.21
    $51.21
    Pack of 25
       $51.21
    Supplier Number: 1215368
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78172-978
    Pore size
    0.45 μm
    New Product
  • 0.1 μm
    90 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $50.46
    $50.46
    Pack of 25
       $50.46
    Supplier Number: 1222230
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78172-488
    Pore size
    0.1 μm
    New Product
  • 0.1 μm
    47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $57.95
    $57.95
    Pack of 100
       $57.95
    Supplier Number: 1214756
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78172-350
    Pore size
    0.1 μm
    New Product
  • 0.22 μm
    90 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $51.47
    $51.47
    Pack of 25
       $51.47
    Supplier Number: 1214920
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78173-310
    Pore size
    0.22 μm
    New Product
  • 0.22 μm
    47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $58.99
    $58.99
    Pack of 100
       $58.99
    Supplier Number: 1214465
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78172-234
    Pore size
    0.22 μm
    New Product
  • 1.2 μm
    47 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $56.96
    $56.96
    Pack of 100
       $56.96
    Supplier Number: 1221008
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78172-756
    Pore size
    1.2 μm