Polyethersulfone (PES) Membranes
À propos de cet article
Polyethersulfone membranes are hydrophilic, with a wide range of pore sizes.
- Hydrophilic format eliminates the need for wetting agents that can potentially interfere with analyses
- Low extractables ensure test results will not be compromised by wetting agents or other extractables
- Low drug and protein binding maximizes recovery of critical drugs or proteins
- Superior burst strength protects the integrity of the membrane under high pressure
- Quality checks, both down and across the membrane, ensure dependable results every time
Polyethersulfone (PES) Filtration Membrane is hydrophilic and cast from pure polyethersulfone polymer. It is designed to remove particulates during general filtration and its low protein and drug binding characteristics make it ideally suited for use in life science applications.
9 Options Available Below
Product Details & Documents
Polyethersulfone membranes are hydrophilic, with a wide range of pore sizes.
- Hydrophilic format eliminates the need for wetting agents that can potentially interfere with analyses
- Low extractables ensure test results will not be compromised by wetting agents or other extractables
- Low drug and protein binding maximizes recovery of critical drugs or proteins
- Superior burst strength protects the integrity of the membrane under high pressure
- Quality checks, both down and across the membrane, ensure dependable results every time
Polyethersulfone (PES) Filtration Membrane is hydrophilic and cast from pure polyethersulfone polymer. It is designed to remove particulates during general filtration and its low protein and drug binding characteristics make it ideally suited for use in life science applications.
Specifications for Product Family
Spécifications
- Catalog No:
- 78173-448
- 78173-644
- 78173-652
- 78172-978
- 78172-488
- 78172-350
- 78173-310
- 78172-234
- 78172-756
- Pore size:
- 0.22 μm
- 0.45 μm
- 1.2 μm
- 0.45 μm
- 0.1 μm
- 0.1 μm
- 0.22 μm
- 0.22 μm
- 1.2 μm
Spécifications
Product Family Options
Informations sur le produit
- Taille de poreØAvailabilityPrice
- Supplier Number: 1214193Spécifications:New Product
More
Specifications
Cat. No.78173-448Pore size0.22 μmNew Product - Supplier Number: 1214532Spécifications:New Product
More
Specifications
Cat. No.78173-644Pore size0.45 μmNew Product - Supplier Number: 1222267Spécifications:New Product
More
Specifications
Cat. No.78173-652Pore size1.2 μmNew Product - Supplier Number: 1215368Spécifications:New Product
More
Specifications
Cat. No.78172-978Pore size0.45 μmNew Product - Supplier Number: 1222230Spécifications:New Product
More
Specifications
Cat. No.78172-488Pore size0.1 μmNew Product - Supplier Number: 1214756Spécifications:New Product
More
Specifications
Cat. No.78172-350Pore size0.1 μmNew Product - Supplier Number: 1214920Spécifications:New Product
More
Specifications
Cat. No.78173-310Pore size0.22 μmNew Product - Supplier Number: 1214465Spécifications:New Product
More
Specifications
Cat. No.78172-234Pore size0.22 μmNew Product - Supplier Number: 1221008Spécifications:New Product
More
Specifications
Cat. No.78172-756Pore size1.2 μmNew Product