Quartz Microfiber Filter
Quartz Microfiber Filter
New Product
Quartz Microfiber Filter
New Product
Certificates
Quartz Microfiber Filter
12 Options Available Below
Product Details & Documents
Quartz microfiber filters, D0QF1 standard grade, binder-free.
  • Excellent retention of very fine particles
  • Exceptional chemical and thermal resistance
  • Excellent weight and dimensional stability with lowest trace metal content
  • High permeation enables large volume of air to pass through
  • Higher temperature stability than glass microfiber filters; up to 900 °C
  • Excellent chemical stability with practically no filter-mass loss in the presence of acid gases
GVS quartz microfiber filters are made with pure quartz microfibers and are free of binders or additives of any kind. These filters have retention, loading, and air permeability features similar to those of glass microfiber filters. However, since they have greater chemical resistance at high temperatures, they can be used in environments where extreme conditions are present, replacing the glass microfiber filters in such cases.
Product Family Options
Product Information
  • Grade
    Ø
    Épaisseur
    Weight
    Availability
    Price
  • Standard
    25 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $150.60
    $150.60
    Pack of 100
       $150.60
    Supplier Number: FP025D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    150 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $2,677.15
    $2,677.15
    Pack of 100
       $2,677.15
    Supplier Number: FP150D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    47 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $231.54
    $231.54
    Pack of 100
       $231.54
    Supplier Number: FP047D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    203 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $6,889.18
    $6,889.18
    Pack of 100
       $6,889.18
    Supplier Number: FP203R0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    70 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $683.90
    $683.90
    Pack of 100
       $683.90
    Supplier Number: FP070D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    110 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,430.42
    $1,430.42
    Pack of 100
       $1,430.42
    Supplier Number: FP110D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    50 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $365.16
    $365.16
    Pack of 100
       $365.16
    Supplier Number: FP050D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    55 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $440.46
    $440.46
    Pack of 100
       $440.46
    Supplier Number: FP055D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    90 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $914.23
    $914.23
    Pack of 100
       $914.23
    Supplier Number: FP090D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    120 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,784.32
    $1,784.32
    Pack of 100
       $1,784.32
    Supplier Number: FP120D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    37 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $176.13
    $176.13
    Pack of 100
       $176.13
    Supplier Number: FP037D0QF1QUFC01
    Specifications:
    New Product
    New Product
  • Standard
    125 mm
    440 μm
    85 g/m²
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,727.70
    $1,727.70
    Pack of 100
       $1,727.70
    Supplier Number: FP125D0QF1QUFC01
    Specifications:
    New Product