Perforated Bench
Perforated bench, stainless steel, 36×12×18"
78469-858
New Product
: REY1840-1236
All-welded, hygienic design resists wear and is easy to clean. Ideal for demanding environments.
- Extremely rugged for long-term performance
- Hygienic design features smooth stainless steel surfaces for effortless cleaning
- Perforated top improves cleanliness and airflow in critical areas
Extremely durable bench is made from 304 brushed stainless steel.
- Désignation:Perforated bench, stainless steel
- W×D×H:36×12×18"