Wall Mount Document Holder
Wall mount document holder, Stainless steel, 12.2W×4D×12"H
Catalog # 78469-884
New Product
Supplier: REYTEK EQUIPMENT
Supplier #: REY1783
Jump to:
Product Details & Documents
Keep cleanroom paperwork organized with this stainless steel document holder. Made from 304 brushed stainless steel with all-welded construction, it’s durable, hygienic, and easy to clean. Wall-mounted for space efficiency, ensuring documents stay secure and accessible in controlled environments.
- Rugged and reliable material is extremely durable for long-term use
- Hygienic design features smooth stainless steel surfaces for effortless cleaning
- Space-saving wall mount design is ideal for cleanrooms and production areas
Perfect for storing SOPs, logs, and essential documentation in gowning rooms and critical environments.
Specifications
- Désignation:Wall mount document holder
- Dimensions:12.2W×4D×12"H
- Matière:Stainless steel