Sodium acetate Min. 98.5% (mass/mass)
20 kg
  77974-671
New Product
 :  198-01077
Sodium acetate Min. 98.5% (mass/mass)
  77974-671
New Product
 :  127-09-3
Sodium acetate Min. 98.5% (mass/mass)
20 kg
 :  127-09-3
$494.73
Each(1/Itm/Bx)
Sodium Acetate is a white, crystalline or crystalline powder with high solubility in water and ethanol. It maintains a pH of 7.5 to 9.0 at 50 g/L concentration. It has high purity and is compliant with JIS standards with additional tests. Used for pH buffer, antifreeze agent, dye, pharmaceutical research agent, color fixative, mordant, photographic chemical, and has a wide range of uses.
 : For research use only (RUO).
 :  Acétate de sodium
  •  :127-09-3
  •  :CH₃COONa
  •  :82.03 g/mol
  •  :Température ambiante
  • Taille:
    20 kg