Coat Hook Bars
New Product
Supplier: REYTEK EQUIPMENT
Jump to:
À propos de cet article
Optimize cleanroom gowning areas with these wall-mounted, all-welded 304 stainless steel coat hook bars. Available with six or twelve durable hooks, they are hygienic, easy to clean, and built for controlled environments.
- 6 or 12 hooks Premium 304 brushed stainless steel provides corrosion resistance Hygienic and rugged material features smooth surfaces for effortless cleaning Space-saving wall mount design is ideal for gowning rooms and production areas
Perfect for lab coats, coveralls, and other cleanroom apparel, ensuring a neat and professional workspace.
2 Options Available Below
Product Details & Documents
Optimize cleanroom gowning areas with these wall-mounted, all-welded 304 stainless steel coat hook bars. Available with six or twelve durable hooks, they are hygienic, easy to clean, and built for controlled environments.
- 6 or 12 hooks Premium 304 brushed stainless steel provides corrosion resistance Hygienic and rugged material features smooth surfaces for effortless cleaning Space-saving wall mount design is ideal for gowning rooms and production areas
Perfect for lab coats, coveralls, and other cleanroom apparel, ensuring a neat and professional workspace.
Product Family Options
Informations sur le produit
- DésignationMatièreLargeurProfondeurHauteurAvailabilityPrice
- Supplier Number: REY1815-006Spécifications:New ProductNew Product
- Supplier Number: REY1815-012Spécifications:New ProductNew Product