Single Bottle Holder
Single bottle holder, stainless steel, 4.7W×3.5D×5.0"H
78469-848
New Product
Supplier #: REY1779
Keep liquids organized with this stainless steel single bottle holder. Made from 304 brushed stainless steel with all-welded construction, it’s durable, hygienic, and easy to clean. Wall-mounted for space efficiency, it is ideal for cleanrooms and controlled environments.
- Holds one bottle (max. diameter 3.48″) Rugged and reliable material is extremely durable for long-term use Hygienic design features a smooth stainless steel surfaces for effortless cleaning Space-saving wall mount design
Perfect for dispensing cleaning solutions or sanitizers in gowning rooms and critical environments.
- Désignation:Single bottle holder, stainless steel
- Dimensions:4.7W×3.5D×5.0"H