Vernier® Go Direct™ Sensor Brackets
Go Direct™ Sensor bracket
Catalog # CA470359-986
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: GDX-MOUNT
Vernier® Go Direct™ Sensor Brackets
Catalog # CA470359-986
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
A convenient, sturdy way to position and hold your sensors in the lab setting.
- Fits most Go Direct box-shaped sensors
Attach the bracket to our accessory rod or any tripod with a standard camera mount to hold a sensor still and stable for repetitive measurements.
Spécifications
- Désignation:Go Direct™ Sensor bracket