Vernier® Go Direct™ Sensor Brackets | Avantor
Vernier® Go Direct™ Sensor Brackets
Go Direct™ Sensor bracket
Catalog # CA470359-986
New Product
Fournisseur #:  GDX-MOUNT
Vernier® Go Direct™ Sensor Brackets
Catalog # CA470359-986
New Product
Vernier® Go Direct™ Sensor Brackets
Go Direct™ Sensor bracket
Price
$11.09
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
A convenient, sturdy way to position and hold your sensors in the lab setting.
  • Fits most Go Direct box-shaped sensors
Attach the bracket to our accessory rod or any tripod with a standard camera mount to hold a sensor still and stable for repetitive measurements.
Spécifications
  • Désignation:
    Go Direct™ Sensor bracket