ScreenFect™ A | Avantor
ScreenFect™ A
1 ml
Catalog # 77974-908
New Product
Fournisseur #:  299-73203
ScreenFect™ A
Catalog # 77974-908
New Product
CAS Number:  
ScreenFect™ A
1 ml
CAS Number:  
Price
$639.08
Log in pour la disponibilité
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
ScreenFect™ A is a low-toxicity transfection reagent made of novel cationic lipids, effective for DNA and siRNA delivery across various eukaryotic cell types - including common cell lines, stem cells, blood cells, microglia, and primary cultures. It can be directly added to media containing antibiotics and serum without needing medium change post-transfection, and contains no harmful substances.
  • High transfection efficiency
  • Non-hazardous
  • Includes dilution buffer
  • Easy to use
  • Suitable for various mammalian cell lines
Caution: For research use only (RUO).
Spécifications
Spécifications
  • Taille:
    1 ml
  • Cat. No.:
    77974-908
  • Storage temperature:
    Refrigerated (at 2...10 °C storage)