Anti-DCPS Rabbit Polyclonal Antibody
100 µl
Catalog # 77942-005
New Product
Supplier: Sino Biological
Supplier #: 204099-T44-100
Jump to:
Product Details & Documents
Produced in rabbits immunized with E. coli-derived Human DCPS fragment, and purified by antigen affinity chromatography.
Type: Primary
Antigen: DCPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Taille:100 µl
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Protein A and antigen affinity
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 °C. Avoid repeated freeze-thaw cycles.
- Host:Rabbit
- Shipping temperature:Ambient
- Antigen name:decapping enzyme, scavenger
- Cat. No.:77942-005
- Reactivity:Human
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:DCPS
- Storage buffer:PBS, pH7.0 with 0.03% Proclin300



