Anti-KRT2 Rabbit Polyclonal Antibody
100 µl
Catalog # 77941-211
New Product
Supplier: Sino Biological
Fournisseur #: 105446-T12-100
Jump to:
Product Details & Documents
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human KRT2/Cytokeratin2, and purified by antigen affinity chromatography.
Type: Primary
Antigen: KRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Taille:100 µl
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Protein A and antigen affinity
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 °C. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Host:Rabbit
- Shipping temperature:Ambient
- Antigen name:keratin 2
- Cat. No.:77941-211
- Reactivity:Human
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:KRT2
- Storage buffer:0.2 μm filtered solution in PBS
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



