Consistometer
Consistometer
Catalog # 470360-236
New Product
Supplier: United Scientific Supplies
Supplier #: CONS01
Jump to:
Product Details & Documents
Determines consistency of almost any viscous substance.
Polished stainless steel construction
- Easy to read 0.5 cm engraved increments
- Spring-loaded gate with instantaneous release
- Two leveling screws for angle adjustment
- Includes a spirit level for precision
This consistometer is the ideal tool to measure consistency and flow rate of viscous substances. The rate of sample flow is measured by observing how far it travels during a specific time interval. A consistometer is commonly used to test consistency of products such as sauces, jams/jellies, dressings, paint, and chemicals.
The consistometer is made of polished stainless steel construction with easy to read 0.5 cm increments. The spring-loaded gate holds about 75 ml of sample. Two leveling screws adjust the angle, and a spirit level helps with precision.
Spécifications
- Désignation:Consistometer