Lead 99.95%, rod, 30 mm
À propos de cet article
As part of our extensive range of rods for industry and research, we offer Lead Rods in a huge choice of 107 variations. Lead is known for its softness, density, high malleability and ductility, as well as being resistant to corrosion and acid. These rods, with their high density, find applications as ballast in ships or counterweights. The malleability of Lead Rods also makes them suitable for radiation shielding applications. With our wide selection of product variations, you’ll find the right rod dimensions to fit your specific application.
- Formule:Pb
11 Options Available Below
Product Details & Documents
As part of our extensive range of rods for industry and research, we offer Lead Rods in a huge choice of 107 variations. Lead is known for its softness, density, high malleability and ductility, as well as being resistant to corrosion and acid. These rods, with their high density, find applications as ballast in ships or counterweights. The malleability of Lead Rods also makes them suitable for radiation shielding applications. With our wide selection of product variations, you’ll find the right rod dimensions to fit your specific application.
- Formule:Pb
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: 1000050488Spécifications:New ProductNew Product
- Supplier Number: 1000050787Spécifications:New ProductNew Product
- Supplier Number: 1000050949Spécifications:New ProductNew Product
- Supplier Number: 1000051126Spécifications:New ProductNew Product
- Supplier Number: 1000165529Spécifications:New ProductNew Product
- Supplier Number: 1000165503Spécifications:New ProductNew Product
- Supplier Number: 1000165507Spécifications:New ProductNew Product
- Supplier Number: 1000165514Spécifications:New ProductNew Product
- Supplier Number: 1000165519Spécifications:New ProductNew Product
- Supplier Number: 1000165526Spécifications:New ProductNew Product
- Supplier Number: 1000196344Spécifications:New ProductNew Product