Lead 99.95%, rod, 30 mm
New Product
Supplier:   Goodfellow
Lead 99.95%, rod, 30 mm
Supplier:  Goodfellow
New Product
Certificates
Lead 99.95%, rod, 30 mm
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
As part of our extensive range of rods for industry and research, we offer Lead Rods in a huge choice of 107 variations. Lead is known for its softness, density, high malleability and ductility, as well as being resistant to corrosion and acid. These rods, with their high density, find applications as ballast in ships or counterweights. The malleability of Lead Rods also makes them suitable for radiation shielding applications. With our wide selection of product variations, you’ll find the right rod dimensions to fit your specific application.
  • Formule:Pb
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • Length 10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $255.67
    $255.67
    Each
       $255.67
    Supplier Number: 1000050488
    Spécifications:
    New Product
    New Product
  • Length 20 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $274.05
    $274.05
    Each
       $274.05
    Supplier Number: 1000050787
    Spécifications:
    New Product
    New Product
  • Length 30 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.60
    $290.60
    Each
       $290.60
    Supplier Number: 1000050949
    Spécifications:
    New Product
    New Product
  • Length 50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $320.74
    $320.74
    Each
       $320.74
    Supplier Number: 1000051126
    Spécifications:
    New Product
    New Product
  • Length 500 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $797.89
    $797.89
    Each
       $797.89
    Supplier Number: 1000165529
    Spécifications:
    New Product
    New Product
  • Length 1000 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,218.13
    $1,218.13
    Each
       $1,218.13
    Supplier Number: 1000165503
    Spécifications:
    New Product
    New Product
  • Length 100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $387.34
    $387.34
    Each
       $387.34
    Supplier Number: 1000165507
    Spécifications:
    New Product
    New Product
  • Length 200 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $503.30
    $503.30
    Each
       $503.30
    Supplier Number: 1000165514
    Spécifications:
    New Product
    New Product
  • Length 300 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $607.68
    $607.68
    Each
       $607.68
    Supplier Number: 1000165519
    Spécifications:
    New Product
    New Product
  • Length 400 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $705.19
    $705.19
    Each
       $705.19
    Supplier Number: 1000165526
    Spécifications:
    New Product
    New Product
  • Length 580 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $869.39
    $869.39
    Each
       $869.39
    Supplier Number: 1000196344
    Spécifications:
    New Product