LaMotte 1762 DO Replacement Sensor Kit | Avantor
LaMotte 1762 DO Replacement Sensor Kit
1762 DO replacement sensor kit
Catalog # 470360-156
New Product
Supplier:   LaMotte
Fournisseur #:  1762
LaMotte 1762 DO Replacement Sensor Kit
Catalog # 470360-156
Supplier:  LaMotte
New Product
LaMotte 1762 DO Replacement Sensor Kit
1762 DO replacement sensor kit
Price
$489.99
Log in pour la disponibilité
Each
Product Details & Documents
Keep your DO meter performing like new. Ensure accurate, reliable results for every classroom experiment.
  • Restores full dissolved oxygen testing capability to the LaMotte 1761 TRACER, keeping this device performing like new
  • All-in-one replacement package includes a pre-assembled DO sensor, membrane cap, and electrolyte solution for quick, hassle-free installation
  • Factory-matched components ensure optimal compatibility and accuracy with the existing TRACER unit
  • User-replaceable design reduces downtime and eliminates the need for factory service, keeping testing schedules on track
  • Durable construction maintains long-term reliability, even with frequent field or lab use
  • Cost-effective maintenance solution extends the lifespan of dissolved oxygen meters without replacing the entire instrument
The LaMotte 1762 DO Replacement Sensor Kit restores the 1761 TRACER dissolved oxygen meter’s accuracy and reliability. This all-in-one kit includes the sensor, membrane cap, and electrolyte solution for quick, easy replacement. Perfect for educators and environmental professionals, it ensures precise DO measurements for classroom labs, fieldwork, and water quality monitoring. Extend your meter’s life with this cost-effective, durable sensor kit.
Spécifications
  • Désignation:
    1762 DO replacement sensor kit