Sacs pour appareils
Compact, FDA compliant, LDPE/Nylon clear blend device liner with a sealable top. Available in sterile and non sterile formats, the device liner is designed specifically for use in clean and controlled environments.
FDA compliant materials; made from a clear LDPE/Nylon laminate (75%/25%)
- Sterile and irradiated version achieves a 10⁻⁶ Sterility Assurance Level, compliant with AAMI guidelines
- Withstands autoclaving, even with liquid contents
- Chemical compatibility: resistant to standard disinfectants and cleaning solutions
- Sealable top ensures secure containment and protection
- Lot and part numbers printed on each package and case for traceability
- 36 month shelf life for long-term usability
- Eco-friendly packaging; ECT-rated case is made from recycled materials
- Animal-free manufacturing; no raw materials of animal or human origin used
The Device Liner is a precision-engineered solution designed to safeguard sensitive electronics and instruments within cleanroom and controlled environments. Manufactured at a Micronova ISO 9001-certified facility, this liner combines regulatory compliance, material integrity, and user-centric design to meet the rigorous demands of pharmaceutical, biotech, and electronics industries.
Crafted from a high-performance blend of FDA-compliant LDPE (75%) and Nylon (25%), the liner offers a clear, durable barrier with a sealable top for secure containment. The Device Liner is available in both sterile and non sterile formats.
The sterile Device Liner is irradiated to a minimum dose of 25 kGy, ensuring a 10⁻⁶ Sterility Assurance Level in accordance with AAMI sterilization guidelines. With a 36 month shelf life, full traceability, and a BSE/TSE-free manufacturing process, both the sterile and non sterile Device Liner delivers peace of mind and operational efficiency in environments where sterility and precision are paramount.
Compact, FDA compliant, LDPE/Nylon clear blend device liner with a sealable top. Available in sterile and non sterile formats, the device liner is designed specifically for use in clean and controlled environments.
FDA compliant materials; made from a clear LDPE/Nylon laminate (75%/25%)
- Sterile and irradiated version achieves a 10⁻⁶ Sterility Assurance Level, compliant with AAMI guidelines
- Withstands autoclaving, even with liquid contents
- Chemical compatibility: resistant to standard disinfectants and cleaning solutions
- Sealable top ensures secure containment and protection
- Lot and part numbers printed on each package and case for traceability
- 36 month shelf life for long-term usability
- Eco-friendly packaging; ECT-rated case is made from recycled materials
- Animal-free manufacturing; no raw materials of animal or human origin used
The Device Liner is a precision-engineered solution designed to safeguard sensitive electronics and instruments within cleanroom and controlled environments. Manufactured at a Micronova ISO 9001-certified facility, this liner combines regulatory compliance, material integrity, and user-centric design to meet the rigorous demands of pharmaceutical, biotech, and electronics industries.
Crafted from a high-performance blend of FDA-compliant LDPE (75%) and Nylon (25%), the liner offers a clear, durable barrier with a sealable top for secure containment. The Device Liner is available in both sterile and non sterile formats.
The sterile Device Liner is irradiated to a minimum dose of 25 kGy, ensuring a 10⁻⁶ Sterility Assurance Level in accordance with AAMI sterilization guidelines. With a 36 month shelf life, full traceability, and a BSE/TSE-free manufacturing process, both the sterile and non sterile Device Liner delivers peace of mind and operational efficiency in environments where sterility and precision are paramount.
Product Family Options
- DésignationW×L
- : BKPH-58New ProductNew Product
- : BKPH-58IRBNew ProductNew Product
- Laptop liner, clear LDPE, 75% Nylon/25% laminate, non sterile18×22"Case of 50$675.09: BKPL-1822New ProductNew Product
- Laptop liner, clear LDPE, 75% Nylon/25% laminate, sterile18×22"Case of 50$746.91: BKPL-1822IRBNew ProductNew Product
- Tablet liner, clear LDPE, 75% Nylon/25% laminate, non sterile12×16"Case of 50$574.55: BKPT-1216New ProductNew Product
- Tablet liner, clear LDPE, 75% Nylon/25% laminate, sterile12×16"Case of 50$646.36: BKPT-1216IRBNew ProductNew Product