Sacs pour appareils
New Product
 :   Micronova
Sacs pour appareils
 :  Micronova
New Product
Sacs pour appareils
Compact, FDA compliant, LDPE/Nylon clear blend device liner with a sealable top. Available in sterile and non sterile formats, the device liner is designed specifically for use in clean and controlled environments.
  • FDA compliant materials; made from a clear LDPE/Nylon laminate (75%/25%)
  • Sterile and irradiated version achieves a 10⁻⁶ Sterility Assurance Level, compliant with AAMI guidelines
  • Withstands autoclaving, even with liquid contents
  • Chemical compatibility: resistant to standard disinfectants and cleaning solutions
  • Sealable top ensures secure containment and protection
  • Lot and part numbers printed on each package and case for traceability
  • 36 month shelf life for long-term usability
  • Eco-friendly packaging; ECT-rated case is made from recycled materials
  • Animal-free manufacturing; no raw materials of animal or human origin used
The Device Liner is a precision-engineered solution designed to safeguard sensitive electronics and instruments within cleanroom and controlled environments. Manufactured at a Micronova ISO 9001-certified facility, this liner combines regulatory compliance, material integrity, and user-centric design to meet the rigorous demands of pharmaceutical, biotech, and electronics industries.
Crafted from a high-performance blend of FDA-compliant LDPE (75%) and Nylon (25%), the liner offers a clear, durable barrier with a sealable top for secure containment. The Device Liner is available in both sterile and non sterile formats.
The sterile Device Liner is irradiated to a minimum dose of 25 kGy, ensuring a 10⁻⁶ Sterility Assurance Level in accordance with AAMI sterilization guidelines. With a 36 month shelf life, full traceability, and a BSE/TSE-free manufacturing process, both the sterile and non sterile Device Liner delivers peace of mind and operational efficiency in environments where sterility and precision are paramount.
Product Family Options
  • Désignation
    W×L
  • Non sterile device liner
    5×8"
    Case of 50
    Case of 50 - $367.71
    $367.71
    Case of 50
       $367.71
     : BKPH-58
    New Product
    New Product
  • Sterile device liner
    5×8"
    Case of 50
    Case of 50 - $441.25
    $441.25
    Case of 50
       $441.25
     : BKPH-58IRB
    New Product
    New Product
  • Laptop liner, clear LDPE, 75% Nylon/25% laminate, non sterile
    18×22"
    Case of 50
    Case of 50 - $675.09
    $675.09
    Case of 50
       $675.09
     : BKPL-1822
    New Product
    New Product
  • Laptop liner, clear LDPE, 75% Nylon/25% laminate, sterile
    18×22"
    Case of 50
    Case of 50 - $746.91
    $746.91
    Case of 50
       $746.91
     : BKPL-1822IRB
    New Product
    New Product
  • Tablet liner, clear LDPE, 75% Nylon/25% laminate, non sterile
    12×16"
    Case of 50
    Case of 50 - $574.55
    $574.55
    Case of 50
       $574.55
     : BKPT-1216
    New Product
    New Product
  • Tablet liner, clear LDPE, 75% Nylon/25% laminate, sterile
    12×16"
    Case of 50
    Case of 50 - $646.36
    $646.36
    Case of 50
       $646.36
     : BKPT-1216IRB