Plain Microscope Slides
Plain Microscope Slides
New Product
Plain Microscope Slides
Plain Microscope Slides
Four E's USA microscope slides are made from high-quality soda glass or super white glass.
  • Pre-cleaned, ready to use
  • Grounded edges, 45° corners
  • Autoclavable
Suitable for traditional identification labeling, these plain slides are manufactured using an advanced grinding process to ensure uniform pieces and consistency in flatness and smoothness. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.
 : Fragile: Handle with care.
Product Family Options
  • Désignation
    Matière
    L×l
    Packaging
  • Plain slides, grounded edges, 45°
    Soda glass
    75×25 mm
    72/box, 20 boxes/carton
    Case of 1
    Case of 1 - $257.11
    $257.11
    Case of 1
       $257.11
     : SLD145110A
    New Product
    New Product
  • Plain slides, grounded edges, 45°
    Super white glass
    75×25 mm
    72/box, 20 boxes/carton
    Case of 1
    Case of 1 - $354.04
    $354.04
    Case of 1
       $354.04
     : SLD245110A