Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Bromothymol blue indicateur de pH

New Product
Supplier:   Abcam
Bromothymol blue indicateur de pH
Supplier:  Abcam
New Product
Certificates

2 Options Available Below

Product Details & Documents

pH indicator for weak acids and bases (pH range from 6.0 (yellow) to 7.6 (blue)). Also used as a stain to define cell walls or nuclei under the microscope.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-59-5
  • Molecular Weight:624.38 g/mol
Specifications for Product Family

Résultats des tests de spécification

  • Constituents::
    100% Bromothymol blue free acid

Spécifications

Product Family Options

Informations sur le produit

  • Taille
    Availability
    Price
  • 25 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (25g)
    Each (25g) - $512.00
    $512.00
    Each (25g)
       $512.00
     

    Supplier Number: AB146293-25G
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB146293-25G
    New Product
  • 5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5g)
    Each (5g) - $144.00
    $144.00
    Each (5g)
       $144.00
     

    Supplier Number: AB146293-5G
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB146293-5G
    New Product