Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Nocardamine ≥95% Antibiotic agent

New Product
 :   Abcam
Nocardamine ≥95% Antibiotic agent
 :  Abcam
New Product
 
Nocardamine ≥95% Antibiotic agent

 

 

Antibiotic agent. Siderophore iron chelator. Induces morphological changes in insect cells. Shows weak antimicrobial against Gram-positive bacteria.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  •  :26605-16-3
  •  :600.7 g/mol
 

 

  • Constituents::
    100% Nocardamine
Product Family Options

 

  • Taille
     
     
  • 1 mg
       
       
    Each (1mg)
    Each (1mg) - $359.00
    $359.00
    Each (1mg)
       $359.00
     

     : AB144842-1MG
     
     
    New Product
    New Product
  • 5 mg
       
       
    Each (5mg)
    Each (5mg) - $1,340.00
    $1,340.00
    Each (5mg)
       $1,340.00
     

     : AB144842-5MG
     
     
    New Product
    New Product