DAPT ≥99% gamma-Secretase inhibitor
À propos de cet article
γ-Secretase inhibitor. Inhibits Notch signalling in vitro. Inhibits total Aβ and Aβ42 production in human primary cultures (IC₅₀ values are 115 and 200 nM, respectively) with no effect on APPα and APPβ levels. Blood-brain barrier permeable. 3D Growth matrix component and component of cerebral organoid differentiation media.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:208255-80-5
- Molecular Weight:432.5 g/mol
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
γ-Secretase inhibitor. Inhibits Notch signalling in vitro. Inhibits total Aβ and Aβ42 production in human primary cultures (IC₅₀ values are 115 and 200 nM, respectively) with no effect on APPα and APPβ levels. Blood-brain barrier permeable. 3D Growth matrix component and component of cerebral organoid differentiation media.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:208255-80-5
- Molecular Weight:432.5 g/mol
Specifications for Product Family
Résultats des tests de spécification
- Constituents::100% DAPT
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: AB120633-50MGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:New ProductNew Product
- Supplier Number: AB120633-10MGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:New ProductNew Product



