Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VU0155041 ≥99% positive allosteric mGluR4 agonist

VU0155041 ≥99% positive allosteric mGluR4 agonist

New Product
Supplier:   Abcam
VU0155041 ≥99% positive allosteric mGluR4 agonist
Supplier:  Abcam
New Product
Certificates
VU0155041 ≥99% positive allosteric mGluR4 agonist

2 Options Available Below

Product Details & Documents

Positive allosteric selective modulator at mGluR4 (EC50 values are 798 and 693 nM at human and rat mGluR4 receptors, respectively). It has advantages over PHCCC (ab120043) as it is more potent. Shows limited blood-brain barrier permeability. Active in behavioural models of Parkinson's disease following icv injection. Available in water-soluble form - please see VU0155041 sodium salt (ab120248).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  cis-2-[[(3,5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid, rel-(1R,2S)-2-((3,5-Dichlorophenyl)carbamoyl)cyclohexane-1-carboxylic acid
  • CAS Number:1093757-42-6
  • Molecular Weight:316.2 g/mol
Specifications for Product Family

Résultats des tests de spécification

  • Constituents::
    100% (1R,2S)-2-[(3,5-dichlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
Product Family Options

Informations sur le produit

  • Taille
    Availability
    Price
  • 10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10mg)
    Each (10mg) - $113.00
    $113.00
    Each (10mg)
       $113.00
    Minimum order quantity is 1

    Supplier Number: AB120244-10MG
    Spécifications:
    New Product
    New Product
  • 50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (50mg)
    Each (50mg) - $318.00
    $318.00
    Each (50mg)
       $318.00
    Minimum order quantity is 1

    Supplier Number: AB120244-50MG
    Spécifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.