Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Linsitinib ≥98%

New Product
 :   Abcam
Linsitinib ≥98%
 :  Abcam
New Product
 
Linsitinib ≥98%

 

 

Potent and selective dual IGF-1R (IC₅₀ = 35 nM in cell-free assay) and Insulin Receptor (IC₅₀ = 75 nM in cell-free assay) kinase inhibitor. Inactive towards Abl, ALK, BTK, EGFR, FGFR1/2 and PKA. Displayed <50% inhibition at 1 μM against a panel of 88 kinases. Blocks IGF-1R autophosphorylation and downstream activation of Akt, ERK1/2 and S6 kinase. Blocks tumor cell line proliferation. Blocks IGF-1R driven tumor xenograft growth.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

 :  Cis-3-(8-Amino-1-(2-phenylquinolin-7-yl)imidazo[1,5-a]pyrazin-3-yl)-1-methylcyclobutan-1-ol
  •  :867160-71-2
  •  :421.5 g/mol
 

 

  • Constituents::
    98% 3-[8-amino-1-(2-phenylquinolin-7-yl)imidazo[1,5-a]pyrazin-3-yl]-1-methylcyclobutan-1-ol
Product Family Options

 

  • Taille
     
     
  • 50 mg
       
       
    Each (50mg)
    Each (50mg) - $1,450.00
    $1,450.00
    Each (50mg)
       $1,450.00
     

     : AB273391-50MG
     
     
    New Product
    New Product
  • 10 mg
       
       
    Each (10mg)
    Each (10mg) - $646.00
    $646.00
    Each (10mg)
       $646.00
     

     : AB273391-10MG
     
     
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.