Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Congo Red ≥98% Amyloid fibril-binding dye

100 mg
Catalog # ABCA_AB145645-100M
New Product
Supplier:   Abcam
 :  AB145645-100MG
Congo Red ≥98% Amyloid fibril-binding dye
Catalog # ABCA_AB145645-100M
Supplier:  Abcam
New Product
CAS Number:  
Congo Red ≥98% Amyloid fibril-binding dye

100 mg

CAS Number:  
Price
$175.00
Log in pour la disponibilité
Each (100mg)
 
Product Details & Documents

Amyloid fibril-binding dye. Caspase inhibitor. Red shifts on aggregation and binding to amyloid fibrils. Blue shifts on binding to cellulose. Shows apple green birefringence under polarized light in the presence of amyloid fibrils. Inhibits Fib-beta A neurotoxicity.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  Direct Red 28
  • Molecular Weight:696.7 g/mol
Spécifications

Spécifications

  • Taille:
    100 mg

Spécifications