Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Loperamide hydrochloride ≥99% mu opioid receptor agonist | Avantor

Loperamide hydrochloride ≥99% mu opioid receptor agonist

1 g
Catalog # ABCA_AB120713-1G
New Product
Supplier:   Abcam
Fournisseur #:  AB120713-1G
Loperamide hydrochloride ≥99% mu opioid receptor agonist
Catalog # ABCA_AB120713-1G
Supplier:  Abcam
New Product
CAS Number:  34552-83-5
Loperamide hydrochloride ≥99% mu opioid receptor agonist

1 g

CAS Number:  34552-83-5
Price
$175.00
Log in pour la disponibilité
Each (1g)
La quantité minimale de commande est de 1
Product Details & Documents

Peripheral selective μ opioid receptor agonist. Ki values are 3, 48 and 1156 nM for opioid receptor subtypes μ, δ and κ, respectively. High doses allow central μ receptor activation. Antidiarrheal, antipruritic and antihyperalgesic agent.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:34552-83-5
  • Molecular Weight:513.5 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    100% Loperamide hydrochloride

Spécifications

  • Taille:
    1 g

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.